Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03678601_g1
          See other LINC00847 GE Assays ›
          Gene Symbol:
          LINC00847
          Gene Name:
          long intergenic non-protein coding RNA 847
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 180830957 - 180835726 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03678601_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00847
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027183.2 1 - 2 276 103
          GenBank mRNA DA361911.1 - 1 - 2 224 103
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AL528925.3 - 1 - 2 194 Chr. 5 - 180831039 - 180834871 [+] on Build GRCh38 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream hepatocellular carcinoma up-regulated EZH2-associated long non-coding RNA HEIH HCCAT2,
          LINC-HEIH,
          LINC00848,
          lncRNA-HEIH
          Anti-sense Chr. 5 - 180829954 - 180831618 on NCBI Build GRCh38
          Downstream ZFP62 zinc finger protein ZFP62 ZET,
          ZNF755
          13.4kb Anti-sense Chr. 5 - 180843580 - 180861327 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04499182 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 14048 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 890059 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 377172 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 175783 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3508717 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5057 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5058 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11035 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55552 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P17419264 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3508717 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3508717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683730 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499163 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15773731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683724 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15773727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15773733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15773728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683724 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499163 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683730 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683751 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15773728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15773727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15773731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15773733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24683744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04499144 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17419264 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499142 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15773742 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499178 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499163 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499168 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499161 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15773740 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499174 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499165 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499153 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499171 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499145 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499170 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499138 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15773739 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499147 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499152 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499166 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15773730 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499156 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15773737 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04499164 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17419264 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1556180_PM_at - -
          Affymetrix Human Genome U219 Array 11755167_s_at - -
          Affymetrix Human Genome U219 Array 11755167_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 1556180_at - -
          Human Genome U133 Plus 2.0 Array 1556180_PM_at - -
          Human Genome U95 Set 34529_PM_at - -
          Human Genome U95 Set 34529_at - -
          Human Genome U95 Set 34530_at - -
          Human Genome U95 Set 34530_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556180_at1 - -
          Human Genome U95 Set 48509_at - -
          Human Genome U95 Set 48509_PM_at - -
          Human Genome U95 Set 43360_PM_s_at - -
          Human Genome U95 Set 43360_s_at - -
          Human Genome U133 Set 226146_at - -
          Human Genome U133 Set 226146_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007490 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007490 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0