Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03678625_m1
          See other LINC01106 GE Assays ›
          Gene Symbol:
          LINC01106 LINC01123
          Gene Name:
          long intergenic non-protein coding RNA 1106
          long intergenic non-protein coding RNA 1123
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.2: 110375109 - 110384536 on Build GRCh38
          Chromosome Location:
          Chr.2: 109986980 - 109995281 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03678625_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01106
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027244.1 1 - 2 221 62
          GenBank mRNA BP341570.1 - 1 - 2 207 62
          Gene Symbol: LINC01123
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046112.1 1 - 2 210 62
          GenBank mRNA DA999847.1 - 1 - 2 141 62
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW675296.1 - 1 - 2 204 Chr. 2 - 109986987 - 109994593 [+] on Build GRCh38 62
          GenBankmRNA BI914615.1 - 1 - 2 114 Chr. 2 - 109987078 - 109995113 [+] on Build GRCh38 62
          GenBankmRNA BI821206.1 - 1 - 2 97 Chr. 2 - 109987093 - 109994923 [+] on Build GRCh38 62
          GenBankmRNA DA353627.1 - 1 - 2 139 Chr. 2 - 109987051 - 109994818 [+] on Build GRCh38 62
          GenBankmRNA HY149276.1 - 1 - 2 128 Chr. 2 - 109987063 - 109994745 [+] on Build GRCh38 62
          GenBankmRNA BP199484.1 - 1 - 2 127 Chr. 2 - 109987063 - 109994857 [+] on Build GRCh38 62
          GenBankmRNA BE167578.1 - 1 - 2 196 Chr. 2 - 109986996 - 109994775 [+] on Build GRCh38 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LIMS3-LOC440895 readthrough LIMS3-LOC440895 -18.5kb Sense Chr. 2 - 109898432 - 109968574 on NCBI Build GRCh38
          Downstream platelet endothelial aggregation receptor 1-like LOC105373985 27.5kb Anti-sense Chr. 2 - 110022295 - 110027562 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200008830.hg.1, TC0200013869.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000674.hg.1, TC02002200.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CNV Microarray 2x400K 91120 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 59099 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3261720 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3228609 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P163009 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P11737 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 22208 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55376 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55014 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3261720 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3228609 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3228609 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3261720 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15302671 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15302671 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P11737 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P163009 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P163009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8044348 - -
          Affymetrix Human Gene 1.0 ST Array 8044350 - -
          Affymetrix Human Gene 1.0 ST Array 8054518 - -
          Affymetrix Human Gene 1.0 ST Array 8054514 - -
          Affymetrix Human Gene 1.0 ST Array 8044347 - -
          Affymetrix Human Gene 1.0 ST Array 8054516 - -
          Affymetrix Human Gene 1.1 ST Array 8054513 - -
          Affymetrix Human Gene 1.1 ST Array 8054517 - -
          Affymetrix Human Gene 1.1 ST Array 8044346 - -
          Affymetrix HT HG-U133+ PM Array Plate 235083_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02010653.hg.1 ENST00000419296.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02010653.hg.1 NR_046110.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02010653.hg.1 NR_046111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02010653.hg.1 NR_046112.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02010653.hg.1 TC02000674.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02010652.hg.1 NR_046110.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02010652.hg.1 NR_046111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02010652.hg.1 NR_046112.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02010652.hg.1 TC02000674.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02035618.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035618.hg.1 NR_027244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035618.hg.1 TC02002200.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02035619.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035619.hg.1 NR_027244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035619.hg.1 TC02002200.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02035617.hg.1 NR_027244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035617.hg.1 TC02002200.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02035623.hg.1 NR_027244.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035623.hg.1 TC02002200.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200201463.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201463.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201463.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201463.hg.1 TC0200013869.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201463.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201463.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201463.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201470.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201470.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201470.hg.1 TC0200013869.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201470.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201470.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161760.hg.1 NR_046110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161760.hg.1 NR_046111.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161760.hg.1 NR_046112.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161760.hg.1 TC0200008830.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200161760.hg.1 NR_046110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161760.hg.1 NR_046111.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161760.hg.1 NR_046112.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 ENST00000419296.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 NR_046110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 NR_046111.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 NR_046112.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 TC0200008830.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 ENST00000419296.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 NR_046110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 NR_046111.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161762.hg.1 NR_046112.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201464.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201464.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201464.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201464.hg.1 TC0200013869.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201464.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201464.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201464.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201465.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201465.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201465.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201465.hg.1 TC0200013869.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201465.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201465.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201465.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 ENST00000419296.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 NR_046110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 NR_046111.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 NR_046112.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 TC0200008830.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 ENST00000419296.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 NR_046110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 NR_046111.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200161761.hg.1 NR_046112.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201462.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201462.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201462.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201462.hg.1 TC0200013869.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201462.hg.1 ENST00000436665.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201462.hg.1 ENST00000448359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201462.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201471.hg.1 NR_027244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201471.hg.1 TC0200013869.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201471.hg.1 NR_027244.1 Refseq
          Human Genome U133 Plus 2.0 Array 235083_PM_at - -
          Human Genome U133 Plus 2.0 Array 235083_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235083_at11 - -
          Human Genome U95 Set 65668_at - -
          Human Genome U95 Set 63488_PM_r_at - -
          Human Genome U95 Set 63487_PM_r_at - -
          Human Genome U95 Set 63488_r_at - -
          Human Genome U95 Set 63487_r_at - -
          Human Genome U95 Set 65668_PM_at - -
          Human Genome U95 Set 74103_PM_at - -
          Human Genome U95 Set 74103_at - -
          Human Genome U133 Set 235083_at - -
          Human Genome U133 Set 235083_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A016166 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007493 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007493 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A016166 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 225894 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0