Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03680369_m1
          See other SNHG17 GE Assays ›
          Gene Symbol:
          SNHG17
          Gene Name:
          small nucleolar RNA host gene 17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 38420592 - 38435375 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03680369_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNHG17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC052370.1 - 2 - 4 230 110
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA N39481.1 - 2 - 3 240 Chr. 20 - 38426472 - 38435308 [-] on Build GRCh38 110
          GenBankmRNA BF340988.1 - 2 - 4 226 Chr. 20 - 38420709 - 38435274 [-] on Build GRCh38 110
          GenBankmRNA AA135803.1 - 2 - 3 201 Chr. 20 - 38426424 - 38435273 [-] on Build GRCh38 110
          GenBankmRNA BM562678.1 - 374 Chr. - - [-] on Build 110
          GenBankmRNA AW629655.1 - 1 - 2 139 Chr. 20 - 38421022 - 38434587 [-] on Build GRCh38 110
          GenBankmRNA AW934973.1 - 3 - 4 Chr. 20 - 38421017 - 38435246 [+] on Build GRCh38 110
          GenBankmRNA BM456729.1 - 434 Chr. - - [-] on Build 110
          GenBankmRNA BG106290.1 - 242 Chr. - - [-] on Build 110
          GenBankmRNA BM720553.1 - 2 - 4 260 Chr. 20 - 38425935 - 38435331 [-] on Build GRCh38 110
          GenBankmRNA BG481678.1 - 2 - 3 205 Chr. 20 - 38420597 - 38435277 [-] on Build GRCh38 110
          GenBankmRNA BG481329.1 - 2 - 3 207 Chr. 20 - 38420625 - 38435277 [-] on Build GRCh38 110
          GenBankmRNA BG330525.1 - 2 - 3 268 Chr. 20 - 38420726 - 38435341 [-] on Build GRCh38 110
          GenBankmRNA CB997412.1 - 300 Chr. - - [-] on Build 110
          GenBankmRNA BU500384.1 - 2 - 3 228 Chr. 20 - 38420792 - 38435297 [-] on Build GRCh38 110
          GenBankmRNA DA482729.1 - 2 - 4 284 Chr. 20 - 38425927 - 38435355 [-] on Build GRCh38 110
          GenBankmRNA BE408029.1 - 2 - 4 186 Chr. 20 - 38420877 - 38435258 [-] on Build GRCh38 110
          GenBankmRNA HY008936.1 - 2 - 4 283 Chr. 20 - 38421065 - 38435352 [-] on Build GRCh38 110
          GenBankmRNA BX093504.1 - 2 - 4 200 Chr. 20 - 38421039 - 38435271 [-] on Build GRCh38 110
          GenBankmRNA BG106683.1 - 2 - 3 261 Chr. 20 - 38420653 - 38435333 [-] on Build GRCh38 110
          GenBankmRNA BQ956982.1 - 258 Chr. - - [-] on Build 110
          GenBankmRNA BI836626.1 - 2 - 3 191 Chr. 20 - 38420804 - 38435263 [-] on Build GRCh38 110
          GenBankmRNA HY178299.1 - 2 - 4 307 Chr. 20 - 38426417 - 38435377 [-] on Build GRCh38 110
          GenBankmRNA BF341505.1 - 2 - 3 439 Chr. 20 - 38422289 - 38435341 [-] on Build GRCh38 110
          GenBankmRNA BU153111.1 - 448 Chr. - - [-] on Build 110
          GenBankmRNA BG336608.1 - 277 Chr. - - [-] on Build 109
          GenBankmRNA BE790426.1 - 2 - 4 209 Chr. 20 - 38420633 - 38435281 [-] on Build GRCh38 110

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          H/ACA box 71D
          SNORA71D U71d Sense Chr. 20 - 38433862 - 38433999 on NCBI Build GRCh38
          Downstream small nucleolar RNA host gene 11 SNHG11 C20orf198,
          LINC00101,
          NCRNA00101
          12.0kb Anti-sense Chr. 20 - 38446654 - 38450921 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0