Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03680611_s1
          See other ILF3-AS1 GE Assays ›
          Gene Symbol:
          ILF3-AS1
          Gene Name:
          ILF3 antisense RNA 1 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 10651862 - 10653872 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03680611_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ILF3-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024333.1 1 56 73
          GenBank mRNA DB002247.1 - 1 56 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900009641.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001172.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3601163 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42866 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3601163 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3601163 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107929 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496013 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881922 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881925 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496022 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496011 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881928 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881926 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881924 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496012 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107926 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881930 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496020 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881927 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107927 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881920 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496015 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881923 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107925 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107928 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496018 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496016 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881929 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496009 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881921 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17107928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881925 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107927 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107926 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107925 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881924 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881920 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496009 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107929 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496020 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496012 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496016 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496013 - -
          Agilent Whole Chr array_18 19 21 A_17_P17107928 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496018 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881921 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881929 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496017 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881928 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881923 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881925 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881922 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881926 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496015 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496011 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881930 - -
          Agilent Whole Chr array_18 19 21 A_17_P10881927 - -
          Agilent Whole Chr array_18 19 21 A_17_P31496022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31496022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17107928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10881926 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10881928 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10881925 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17107931 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10881921 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17107929 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10881926 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10881929 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10881922 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17107928 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8034098 - -
          Affymetrix Human Gene 1.1 ST Array 8034097 - -
          Affymetrix HT HG-U133+ PM Array Plate 235014_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19016327.hg.1 TC19001172.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19016327.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178138.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178138.hg.1 TC1900009641.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900178138.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178137.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178137.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178137.hg.1 TC1900009641.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900178137.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178137.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178134.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178134.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178134.hg.1 TC1900009641.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900178134.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178134.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009641.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178135.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178135.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178135.hg.1 TC1900009641.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900178135.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178135.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178136.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178136.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178136.hg.1 TC1900009641.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900178136.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178136.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178133.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178133.hg.1 NR_024333.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900178133.hg.1 TC1900009641.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900178133.hg.1 ENST00000591501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900178133.hg.1 NR_024333.1 Refseq
          Human Genome U133 Plus 2.0 Array 235014_PM_at - -
          Human Genome U133 Plus 2.0 Array 235014_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235014_at2 - -
          Human Genome U95 Set 70917_at - -
          Human Genome U95 Set 75314_PM_at - -
          Human Genome U95 Set 70917_PM_at - -
          Human Genome U95 Set 75314_at - -
          Human Genome U133 Set 235014_at - -
          Human Genome U133 Set 235014_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 230789 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3235597 BC044911.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3235597 AF439744.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3235597 AL832185.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3235597 NR_024333.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235597 BC044911.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235597 AF439744.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235597 AL832185.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235597 NR_024333.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235597 BC044911.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235597 AF439744.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235597 AL832185.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235597 NR_024333.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235597 BC044911.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235597 AF439744.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235597 AL832185.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235597 NR_024333.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6cbc4fbdf-xtbd4:80/100.66.128.131:80.
          git-commit: 26a732473c29a7560faabcf99da408bd94702681
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc