Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000561864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000561864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000561864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028019.hg.1 |
ENST00000397340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028019.hg.1 |
ENST00000538806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028019.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028019.hg.1 |
TC12002128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028021.hg.1 |
ENST00000291374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028021.hg.1 |
ENST00000397340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028021.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028021.hg.1 |
TC12002128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028020.hg.1 |
ENST00000291374.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028020.hg.1 |
ENST00000397340.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028020.hg.1 |
ENST00000538806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028020.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028020.hg.1 |
TC12002128.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
TC1200012435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197251.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
TC1200012435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197253.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197254.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197254.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197254.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197254.hg.1 |
TC1200012435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200197254.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197254.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197254.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000561864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000561864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000561864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012435.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
NR_024457.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
TC1200012435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
ENST00000291374.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
ENST00000567788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200197252.hg.1 |
NR_024457.2 |
Refseq |