Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03681477_m1
          See other CALCOCO1 GE Assays ›
          Gene Symbol:
          CALCOCO1
          Gene Name:
          calcium binding and coiled-coil domain 1
          Gene Aliases:
          Cocoa, PP13275, calphoglin
          Chromosome Location:
          Chr.12: 53711118 - 53727523 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03681477_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CALCOCO1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026554.1 2 - 3 300 95
          GenBank mRNA AK296497.1 - 2 - 3 229 95
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DC390027.1 - 2 - 3 229 Chr. 12 - 53722022 - 53727452 [-] on Build GRCh38 95
          GenBankmRNA DC407608.1 - 2 - 3 229 Chr. 12 - 53722061 - 53727452 [-] on Build GRCh38 95
          GenBankmRNA DC308821.1 - 2 - 3 221 Chr. 12 - 53722046 - 53727443 [-] on Build GRCh38 95
          GenBankmRNA DC306790.1 - 2 - 3 229 Chr. 12 - 53722049 - 53727452 [-] on Build GRCh38 95
          GenBankmRNA BP328880.1 - 2 - 3 234 Chr. 12 - 53722027 - 53727457 [-] on Build GRCh38 95
          GenBankmRNA DC352601.1 - 2 - 3 229 Chr. 12 - 53722060 - 53727452 [-] on Build GRCh38 95
          GenBankmRNA DC344416.1 - 2 - 3 234 Chr. 12 - 53722055 - 53727457 [-] on Build GRCh38 95
          GenBankmRNA DC407630.1 - 2 - 3 229 Chr. 12 - 53722055 - 53727452 [-] on Build GRCh38 95
          GenBankmRNA BP311774.1 - 2 - 3 228 Chr. 12 - 53722022 - 53727452 [-] on Build GRCh38 95
          GenBankmRNA DC408691.1 - 2 - 3 231 Chr. 12 - 53722060 - 53727453 [-] on Build GRCh38 95
          GenBankmRNA DC343823.1 - 2 - 3 229 Chr. 12 - 53722055 - 53727452 [-] on Build GRCh38 95
          GenBankmRNA EL594823.1 - 3 - 4 Chr. 12 - 53721543 - 53725182 [+] on Build GRCh38 95
          GenBankmRNA BP328837.1 - 2 - 3 234 Chr. 12 - 53722031 - 53727457 [-] on Build GRCh38 95
          GenBankmRNA DC339879.1 - 2 - 3 223 Chr. 12 - 53722041 - 53727446 [-] on Build GRCh38 95
          GenBankmRNA DC422955.1 - 2 - 3 223 Chr. 12 - 53722054 - 53727446 [-] on Build GRCh38 95
          GenBankmRNA DC423207.1 - 2 - 3 229 Chr. 12 - 53722058 - 53727452 [-] on Build GRCh38 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATP synthase,
          H+ transporting,
          mitochondrial Fo complex subunit C2 (subunit 9)
          ATP5G2 ATP5A -46.9kb Sense Chr. 12 - 53665160 - 53677546 on NCBI Build GRCh38
          Downstream chondrogenesis-associated transcript CISTR CISTR-ACT,
          CISTRACT,
          re52431
          25.2kb Sense Chr. 12 - 53750447 - 53757034 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK296497.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          signal transduction
          positive regulation of gene expression
          Wnt signaling pathway
          intracellular steroid hormone receptor signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          transcription cis-regulatory region binding
          core promoter sequence-specific DNA binding
          chromatin binding
          transcription coregulator activity
          transcription coactivator activity
          protein binding
          beta-catenin binding
          protein C-terminus binding
          nuclear receptor coactivator activity
          sequence-specific DNA binding
          armadillo repeat domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0