Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03826402_s1
          See other PIH2 GE Assays ›
          Gene Symbol:
          PIH2
          Gene Name:
          pregnancy-induced hypertension syndrome-related protein
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03826402_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIH2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF232217.1 - 1 145 186

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101929759 LOC101929759 -100.3kb Sense Chr. 8 - 69834105 - 69837829 on NCBI Build GRCh38
          Downstream syndecan binding protein pseudogene 2 SDCBPP2 4.5kb Sense Chr. 8 - 69942921 - 69944286 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06198562 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06198548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06198557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06198555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16104255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06198560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26481550 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P918214 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 22415 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P918214 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P918214 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 234426_PM_x_at - -
          Affymetrix HT HG-U133+ PM Array Plate 234420_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0800007911.hg.1 ENST00000623186.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007911.hg.1 ENST00000623186.1 Ensembl
          Human Genome U133 Plus 2.0 Array 234420_at - -
          Human Genome U133 Plus 2.0 Array 234426_x_at - -
          Human Genome U133 Plus 2.0 Array 234420_PM_at - -
          Human Genome U133 Plus 2.0 Array 234426_PM_x_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234426_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234420_at3 - -
          Human Genome U133 Set 234420_at - -
          Human Genome U133 Set 234426_PM_x_at - -
          Human Genome U133 Set 234420_PM_at - -
          Human Genome U133 Set 234426_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1898999 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 5420500 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_108610 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1898999 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1898999 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1898999 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0