Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03831718_s1
          See other MFSD9 GE Assays ›
          Gene Symbol:
          MFSD9
          Gene Name:
          major facilitator superfamily domain containing 9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 102713867 - 102736888 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03831718_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MFSD9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001322080.1 NP_001309009.1 6 5146 97
          NM_001322081.1 NP_001309010.1 6 5047 97
          NM_032718.4 NP_116107.3 6 5030 97
          NR_136173.1 7 5114 97
          NR_136174.1 7 2851 97
          XM_011512005.2 XP_011510307.1 6 5153 97
          XM_011512006.2 XP_011510308.1 6 4970 97
          XM_011512007.2 XP_011510309.1 4 4627 97
          GenBank mRNA CA426861.1 - 1 97
          U79290.1 - 1 1552 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI697733.1 - 1 Chr. 2 - 102714633 - 102715131 [+] on Build GRCh38 97
          GenBankmRNA AI659212.1 - 1 Chr. 2 - 102714630 - 102715044 [+] on Build GRCh38 97
          GenBankmRNA AI041723.1 - 1 Chr. 2 - 102714630 - 102715040 [+] on Build GRCh38 97
          GenBankmRNA AW517278.1 - 1 Chr. 2 - 102714623 - 102715009 [+] on Build GRCh38 97
          GenBankmRNA BE674234.1 - 1 Chr. 2 - 102714703 - 102715125 [+] on Build GRCh38 97
          GenBankmRNA H09540.1 - 1 Chr. 2 - 102714630 - 102715007 [+] on Build GRCh38 97
          GenBankmRNA AI804340.1 - 1 Chr. 2 - 102714630 - 102715115 [+] on Build GRCh38 97
          GenBankmRNA BF375129.1 - 1 Chr. 2 - 102714706 - 102715097 [+] on Build GRCh38 97
          GenBankmRNA BF910239.1 - 1 Chr. 2 - 102714729 - 102714932 [+] on Build GRCh38 97
          GenBankmRNA BF909777.1 - 1 Chr. 2 - 102714735 - 102714942 [+] on Build GRCh38 97
          GenBankmRNA BF095024.1 - 1 Chr. 2 - 102714730 - 102714932 [-] on Build GRCh38 97
          GenBankmRNA AA626095.1 - 1 Chr. 2 - 102714632 - 102714964 [+] on Build GRCh38 97
          GenBankmRNA AW474044.1 - 1 Chr. 2 - 102714630 - 102715110 [+] on Build GRCh38 97
          GenBankmRNA BM311581.1 - 1 Chr. 2 - 102714630 - 102715181 [+] on Build GRCh38 97
          GenBankmRNA AI381480.1 - 1 Chr. 2 - 102714630 - 102715089 [+] on Build GRCh38 97
          GenBankmRNA AI280116.1 - 1 Chr. 2 - 102714630 - 102715081 [+] on Build GRCh38 97
          GenBankmRNA AI040212.1 - 1 Chr. 2 - 102714630 - 102715051 [+] on Build GRCh38 97
          GenBankmRNA AI950305.1 - 1 Chr. 2 - 102714623 - 102715095 [+] on Build GRCh38 97
          GenBankmRNA BF111097.1 - 1 Chr. 2 - 102714630 - 102715109 [+] on Build GRCh38 97
          GenBankmRNA AW001194.1 - 1 Chr. 2 - 102714631 - 102715145 [+] on Build GRCh38 97
          GenBankmRNA AW517757.1 - 1 Chr. 2 - 102714627 - 102714933 [+] on Build GRCh38 98
          GenBankmRNA CK821568.1 - 1 Chr. 2 - 102714630 - 102715206 [+] on Build GRCh38 97
          GenBankmRNA AW513816.1 - 1 Chr. 2 - 102714631 - 102715109 [+] on Build GRCh38 97
          GenBankmRNA BP395460.1 - 1 231 Chr. 2 - 102714630 - 102715123 [-] on Build GRCh38 97
          GenBankmRNA BE671364.1 - 1 Chr. 2 - 102714703 - 102715122 [+] on Build GRCh38 97
          GenBankmRNA AI689888.1 - 1 Chr. 2 - 102714633 - 102715022 [+] on Build GRCh38 97
          GenBankmRNA BF223370.1 - 1 Chr. 2 - 102714630 - 102715172 [+] on Build GRCh38 97
          GenBankmRNA AA746270.1 - 1 Chr. 2 - 102714630 - 102715035 [+] on Build GRCh38 97
          GenBankmRNA CK821567.1 - 1 485 Chr. 2 - 102714809 - 102715377 [-] on Build GRCh38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 9 member A2 SLC9A2 NHE2 -3.3kb Anti-sense Chr. 2 - 102619689 - 102711350 on NCBI Build GRCh38
          Downstream transmembrane protein 182 TMEM182 20.5kb Anti-sense Chr. 2 - 102736906 - 102843838 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MFSD9

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane transport
          transporter activity

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 197256 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0