Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03833056_s1
          See other FOXN1 GE Assays ›
          Gene Symbol:
          FOXN1
          Gene Name:
          forkhead box N1
          Gene Aliases:
          FKHL20, RONU, WHN
          Chromosome Location:
          Chr.17: 28506211 - 28538900 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03833056_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FOXN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005258046.3 XP_005258103.1 9 3307 102
          XM_011525358.1 XP_011523660.1 9 3310 102
          XM_011525359.1 XP_011523661.1 9 3293 102
          XM_011525362.1 XP_011523664.1 9 3191 102
          XM_011525367.1 XP_011523669.1 7 3098 102
          XM_011525368.2 XP_011523670.1 7 2727 102
          XM_011525369.2 XP_011523671.1 7 2875 102
          XM_011525370.2 XP_011523672.1 7 2872 102
          XM_017025228.1 XP_016880717.1 9 3322 102
          XM_017025229.1 XP_016880718.1 9 3047 102
          XM_017025230.1 XP_016880719.1 8 2801 102
          GenBank mRNA AF086453.1 - 1 254 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA653683.1 - 1 Chr. 17 - 28538507 - 28538898 [-] on Build GRCh38 102
          GenBankmRNA AI092869.1 - 1 Chr. 17 - 28538467 - 28538896 [-] on Build GRCh38 102
          GenBankmRNA BX111784.1 - 1 254 Chr. 17 - 28538316 - 28538887 [+] on Build GRCh38 102
          GenBankmRNA AW974717.1 - 1 130 Chr. 17 - 28538471 - 28538854 [+] on Build GRCh38 102
          GenBankmRNA HY268961.1 - 1 Chr. 17 - 28538322 - 28538741 [-] on Build GRCh38 102
          GenBankmRNA W80890.1 - 254 Chr. - - [-] on Build 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 13 member 2 SLC13A2 NADC1,
          NaCT,
          NaDC-1,
          SDCT1
          -40.5kb Sense Chr. 17 - 28473358 - 28497781 on NCBI Build GRCh38
          Downstream unc-119 lipid binding chaperone UNC119 HRG4,
          IMD13,
          POC7,
          POC7A
          7.8kb Anti-sense Chr. 17 - 28546707 - 28552633 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          T cell lineage commitment
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          defense response
          epidermis development
          animal organ morphogenesis
          keratinocyte differentiation
          positive regulation of epithelial cell differentiation
          nail development
          T cell homeostasis
          blood vessel morphogenesis
          epithelial cell proliferation
          positive regulation of hair follicle development
          lymphoid lineage cell migration into thymus
          thymus epithelium morphogenesis
          regulation of positive thymic T cell selection
          DNA-binding transcription factor activity, RNA polymerase II-specific
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000577936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000577936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000577936.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000226247.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000577936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000577936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000577936.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 ENST00000579795.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007372.hg.1 NM_003593.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0