Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03834194_s1
          See other CADM3-AS1 GE Assays ›
          Gene Symbol:
          CADM3-AS1
          Gene Name:
          CADM3 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 159195985 - 159202422 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03834194_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CADM3-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037870.1 4 3227 98
          GenBank mRNA AL050219.1 - 1 952 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cell adhesion molecule 3 CADM3 BIgR,
          IGSF4B,
          NECL1,
          Necl-1,
          TSLL1,
          synCAM3
          Anti-sense Chr. 1 - 159171587 - 159203142 on NCBI Build GRCh38
          Downstream atypical chemokine receptor 1 (Duffy blood group) ACKR1 CCBP1,
          CD234,
          DARC,
          Dfy,
          FY,
          GPD,
          GpFy,
          WBCQ1
          7.0kb Anti-sense Chr. 1 - 159204013 - 159206500 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100016088.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003385.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 36960 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3359953 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 217442_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000609696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 ENST00000609696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01052863.hg.1 ENST00000415675.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01052863.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01052863.hg.1 TC01003385.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01052867.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01052867.hg.1 TC01003385.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01052864.hg.1 ENST00000415675.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01052864.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01052864.hg.1 TC01003385.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01052866.hg.1 ENST00000415675.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01052866.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01052866.hg.1 TC01003385.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01052865.hg.1 ENST00000415675.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01052865.hg.1 NR_037870.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01052865.hg.1 TC01003385.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100222181.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222181.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222181.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222181.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222181.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222181.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222181.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222186.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222186.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222186.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222186.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222186.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222186.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222186.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 ENST00000609696.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 ENST00000609696.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222189.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222180.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222180.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222180.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222180.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222180.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222180.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222180.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222188.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222188.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222188.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222188.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222188.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222188.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222188.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222190.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222190.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222190.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222190.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222190.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222190.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222190.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222184.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222184.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222184.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222184.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222184.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222184.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222184.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222187.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222187.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222187.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222187.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222187.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222187.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222187.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000609696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 ENST00000609696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016088.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222191.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222191.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222191.hg.1 NR_037870.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100222191.hg.1 TC0100016088.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100222191.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222191.hg.1 ENST00000415675.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100222191.hg.1 NR_037870.1 Refseq
          Human Genome U133 Plus 2.0 Array 216535_PM_at - -
          Human Genome U133 Plus 2.0 Array 216535_at - -
          Human Genome U133 Plus 2.0 Array 217442_PM_at - -
          Human Genome U133 Plus 2.0 Array 217442_at - -
          Human Genome U133A 2.0 Array 217442_at - -
          Human Genome U133A 2.0 Array 217442_PM_at - -
          Human Genome U133A 2.0 Array 216535_PM_at - -
          Human Genome U133A 2.0 Array 216535_at - -
          AffymetrixPathwork Diagnostics Pathchip pw507602 - -
          AffymetrixPathwork Diagnostics Pathchip pw507189 - -
          Human Genome U95 Set 39287_PM_at - -
          Human Genome U95 Set 39287_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216535_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217442_at4 - -
          Human Genome U133 Set 216535_PM_at - -
          Human Genome U133 Set 216535_at - -
          Human Genome U133 Set 217442_PM_at - -
          Human Genome U133 Set 217442_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053987 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053998 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053994 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053987 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053994 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053998 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3202521 NR_037870.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3202521 AK124069.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0