Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177409.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177409.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177409.hg.1 |
TC1000011395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177409.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177409.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177408.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177408.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177408.hg.1 |
TC1000011395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177408.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177408.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011395.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177407.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177407.hg.1 |
NR_110657.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177407.hg.1 |
TC1000011395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177407.hg.1 |
ENST00000418379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177407.hg.1 |
NR_110657.1 |
Refseq |