Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03846748_s1
          See other LOC100506557 GE Assays ›
          Gene Symbol:
          LOC100506557
          Gene Name:
          uncharacterized LOC100506557
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 157525740 - 157527319 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03846748_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100506557
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK098637.1 - 1 1451 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101927914 LOC101927914 -26.1kb Sense Chr. 7 - 157466231 - 157499716 on NCBI Build GRCh38
          Downstream protein tyrosine phosphatase,
          receptor type N2
          PTPRN2 IA-2beta,
          IAR,
          ICAAR,
          PTPRP,
          R-PTP-N2
          11.7kb Anti-sense Chr. 7 - 157539052 - 158587802 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16047009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16047006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897887 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897888 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16047008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897906 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897889 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167129 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26167132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05897856 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P358063 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 41148 - -
          Agilent Human Genome CGH Microarray 44B A_14_P200796 - -
          Agilent Human Genome CGH Microarray 44B A_14_P117586 - -
          Agilent Human Genome CGH Microarray 44A A_14_P117586 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P358063 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P117586 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P200796 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P117586 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P200796 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P358063 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 15414 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 12790 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 94445 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P117586 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P200796 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8137636 - -
          Affymetrix Human Gene 1.1 ST Array 8137635 - -
          Affymetrix Human Transcriptome Array 2.0 TC0700009894.hg.1 ENST00000624122.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009894.hg.1 ENST00000624122.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1564584_PM_at - -
          Human Genome U133 Plus 2.0 Array 1564584_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564584_at6 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix