Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC034565.1 | - | 1 - 2 | 599 | 95 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | peptidase D | PEPD | PROLIDASE | -33.7kb | Anti-sense | Chr. 19 - 33386949 - 33521893 on NCBI Build GRCh38 |
Downstream | carbohydrate sulfotransferase 8 | CHST8 |
GALNAC4ST1, GalNAc4ST, PSS3 |
64.5kb | Sense | Chr. 19 - 33621955 - 33773509 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |