Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03848412_s1
          See other LINC00641 GE Assays ›
          Gene Symbol:
          LINC00641
          Gene Name:
          long intergenic non-protein coding RNA 641
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 21200079 - 21206900 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03848412_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00641
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038970.1 1 1047 101
          GenBank mRNA BC035145.1 - 1 1050 101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF962167.1 - 1 324 Chr. 14 - 21204402 - 21204774 [-] on Build GRCh38 101
          GenBankmRNA DA319543.1 - 1 117 Chr. 14 - 21204047 - 21204573 [-] on Build GRCh38 101
          GenBankmRNA DB279453.1 - 1 64 Chr. 14 - 21203973 - 21204520 [-] on Build GRCh38 101
          GenBankmRNA CV393152.1 - 1 Chr. 14 - 21204402 - 21204771 [+] on Build GRCh38 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream olfactory receptor family 5 subfamily AU member 1 OR5AU1 OR14-38 -44.1kb Sense Chr. 14 - 21154937 - 21156025 on NCBI Build GRCh38
          Downstream heterogeneous nuclear ribonucleoprotein C (C1/C2) HNRNPC C1,
          C2,
          HNRNP,
          HNRPC,
          SNRPC
          3.6kb Sense Chr. 14 - 21209136 - 21269479 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008624.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000914.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 165041 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 388641 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 326471 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165020 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3543133 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P931030 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 29365 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 48883 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02872029 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3543133 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3543133 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P931030 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P02872029 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02872029 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P931030 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 14837 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 13883 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02872029 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19973854 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02872020 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02872023 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19973845 - -
          Agilent Human 1B Microarray G4111A A_32_P761678 - -
          Agilent Human 1B Microarray G4111A A_32_P346729 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19973845 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02872029 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 240902_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009891.hg.1 NR_038970.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009891.hg.1 TC14000914.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400162185.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400162185.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400162185.hg.1 TC1400008624.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400162185.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400162185.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400162186.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400162186.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400162186.hg.1 TC1400008624.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400162186.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400162186.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555379.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 ENST00000555688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038970.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038971.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008624.hg.1 NR_038971.1 Refseq
          Human Genome U133 Plus 2.0 Array 240902_at - -
          Human Genome U133 Plus 2.0 Array 240902_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240902_at8 - -
          Human Genome U95 Set 62134_PM_at - -
          Human Genome U95 Set 62134_at - -
          Human Genome U95 Set 88306_PM_at - -
          Human Genome U95 Set 91262_PM_at - -
          Human Genome U95 Set 91262_at - -
          Human Genome U95 Set 74355_at - -
          Human Genome U95 Set 74355_PM_at - -
          Human Genome U95 Set 88306_at - -
          Human Genome U133 Set 240902_at - -
          Human Genome U133 Set 240902_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050569 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050568 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050569 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050568 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 703080 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0