Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013450.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013450.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013450.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013450.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013450.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013450.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013450.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013450.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013450.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013450.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013450.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013450.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200502.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200502.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200502.hg.1 |
TC0300013450.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300200502.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200502.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200503.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200503.hg.1 |
NR_046662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200503.hg.1 |
TC0300013450.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300200503.hg.1 |
ENST00000422718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200503.hg.1 |
NR_046662.2 |
Refseq |