Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159216.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159216.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159216.hg.1 |
TC2000008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159216.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159216.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159215.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159215.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159215.hg.1 |
TC2000008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159215.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159215.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008418.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159217.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159217.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159217.hg.1 |
TC2000008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159217.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159217.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159218.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159218.hg.1 |
NR_109868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000159218.hg.1 |
TC2000008418.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000159218.hg.1 |
ENST00000456265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000159218.hg.1 |
NR_109868.1 |
Refseq |