Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03855811_s1
          See other LOC100506031 GE Assays ›
          Gene Symbol:
          LOC100506031
          Gene Name:
          uncharacterized LOC100506031
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03855811_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100506031
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK092372.1 - 1 1945 86
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY070020.1 - 1 359 Chr. 16 - 69324955 - 69325436 [-] on Build GRCh38 86
          GenBankmRNA AI634195.1 - 1 53 Chr. 16 - 69324720 - 69325128 [-] on Build GRCh38 86
          GenBankmRNA DA402524.1 - 1 496 Chr. 16 - 69324998 - 69325572 [-] on Build GRCh38 86
          GenBankmRNA BM837496.1 - 1 284 Chr. 16 - 69324856 - 69325362 [-] on Build GRCh38 86
          GenBankmRNA DB106152.1 - 1 Chr. 16 - 69324762 - 69325316 [+] on Build GRCh38 86
          GenBankmRNA DA621133.1 - 1 242 Chr. 16 - 69324700 - 69325320 [-] on Build GRCh38 86
          GenBankmRNA HY103397.1 - 1 359 Chr. 16 - 69324977 - 69325436 [-] on Build GRCh38 86
          GenBankmRNA DA325863.1 - 1 307 Chr. 16 - 69324820 - 69325384 [-] on Build GRCh38 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream vacuolar protein sorting 4 homolog A VPS4A SKD1,
          SKD1A,
          SKD2,
          VPS4,
          VPS4-1
          Anti-sense Chr. 16 - 69311384 - 69325043 on NCBI Build GRCh38
          Downstream peptide deformylase (mitochondrial) PDF 1.6kb Sense Chr. 16 - 69328621 - 69330595 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 499689 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P218295 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 25893 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03169725 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P218295 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P218295 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03169725 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000564399.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000564399.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000564399.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000254950.12 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000562754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000564399.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000564399.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000564399.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 ENST00000566354.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq
          Affymetrix Human Clariom D Assay TC1600011421.hg.1 NM_013245.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0