Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000444955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000444955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000444955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000450695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000450695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000450695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000464.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000464.hg.1 |
ENST00000435062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000464.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02000464.hg.1 |
TC02000036.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000441014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000444955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000444955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000444955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000450695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000450695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000450695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006597.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
NR_038432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
TC0200006597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000419713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000435062.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
ENST00000457568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200145225.hg.1 |
NR_038432.1 |
Refseq |