Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03860292_s1
          See other LOC100288637 GE Assays ›
          Gene Symbol:
          LOC100288637
          Gene Name:
          OTU deubiquitinase 7A pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 30646115 - 30773009 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03860292_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100288637
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038253.1 6 3827 97
          NR_038254.1 5 3727 97
          NR_038255.1 4 3632 97
          GenBank mRNA AK092283.1 - 1 841 97
          AK095096.1 - 1 1752 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Rho GTPase activating protein 11B ARHGAP11B B'-T,
          FAM7B1
          -130.5kb Sense Chr. 15 - 30626676 - 30638810 on NCBI Build GRCh38
          Downstream putative golgin subfamily A member 8I LOC101930434 18.5kb Sense Chr. 15 - 30798436 - 30802763 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16771218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048743 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048770 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16771041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16771208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16771070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09562473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16771176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30048848 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 128749 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 868 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P57269592 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03000269 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P40346894 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P20198867 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03000269 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03000278 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P20198867 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03000272 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P40346894 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 174544 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 93077 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 66705 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 38138 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 87888 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 88628 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 14037 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12161818 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03000272 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12161158 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03000269 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12164305 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12162073 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12162093 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12162170 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12161548 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20198867 - -
          Agilent Human 1B Microarray G4111A A_32_P370617 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052711 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052710 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052710 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052711 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 231320 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0