Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03868111_s1
          See other TMEM178B GE Assays ›
          Gene Symbol:
          TMEM178B
          Gene Name:
          transmembrane protein 178B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 141073402 - 141511338 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03868111_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMEM178B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195278.1 NP_001182207.1 4 5403 92
          GenBank mRNA AK096481.1 - 1 2749 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA W21855.1 - Chr. - - [-] on Build 92
          GenBankmRNA DB325677.1 - 1 Chr. 7 - 141474868 - 141475444 [-] on Build GRCh38 92
          GenBankmRNA DB316218.1 - 1 Chr. 7 - 141474886 - 141475444 [-] on Build GRCh38 92
          GenBankmRNA AW296660.1 - 1 Chr. 7 - 141475116 - 141475467 [-] on Build GRCh38 92
          GenBankmRNA AV722628.1 - 1 430 Chr. 7 - 141474796 - 141475470 [+] on Build GRCh38 92
          GenBankmRNA BP398440.1 - 1 289 Chr. 7 - 141474937 - 141475436 [+] on Build GRCh38 92
          GenBankmRNA N30405.1 - 1 Chr. 7 - 141475066 - 141475471 [-] on Build GRCh38 92
          GenBankmRNA AI500314.1 - 1 Chr. 7 - 141474973 - 141475445 [-] on Build GRCh38 92
          GenBankmRNA DA334233.1 - 1 309 Chr. 7 - 141474917 - 141475485 [+] on Build GRCh38 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mitochondrial ribosomal protein S33 MRPS33 CGI-139,
          MRP-S33,
          PTD003,
          S33mt
          -457.5kb Anti-sense Chr. 7 - 141006161 - 141014981 on NCBI Build GRCh38
          Downstream acylglycerol kinase AGK CATC5,
          CTRCT38,
          MTDPS10,
          MULK
          75.8kb Sense Chr. 7 - 141551278 - 141655244 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700009386.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000888.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831992 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090759 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090810 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090750 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16025207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090739 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090742 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832027 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090740 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831975 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090768 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090722 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090743 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16025209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16025205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090735 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831966 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832020 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090746 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090748 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16025203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090775 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16025210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090720 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090741 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05831951 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05832031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26090796 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P70245 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 14638 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3226060 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P70245 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P70245 - -
          Agilent Human 1B Microarray G4111A A_32_P490485 - -
          Agilent Human 1B Microarray G4111A A_32_P908572 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07013804.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07013804.hg.1 TC07000888.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07013806.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07013806.hg.1 TC07000888.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07013807.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07013807.hg.1 TC07000888.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07013805.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07013805.hg.1 TC07000888.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 ENST00000563442.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 ENST00000563442.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166589.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166590.hg.1 ENST00000563442.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166590.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166590.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166590.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166590.hg.1 ENST00000563442.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166590.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166590.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166595.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166595.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166595.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166595.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166595.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166595.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166595.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166597.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166597.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166597.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166597.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166597.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166597.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166597.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166598.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166598.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166598.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166598.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166598.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166598.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166598.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166585.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166585.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166585.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166585.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166585.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166585.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166585.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 ENST00000563442.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 ENST00000563442.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166586.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166593.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166593.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166593.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166593.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166593.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166593.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166593.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166596.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166596.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166596.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166596.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166596.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166596.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166596.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166584.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166584.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166584.hg.1 NM_001195278.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700166584.hg.1 TC0700009386.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700166584.hg.1 ENST00000565468.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166584.hg.1 ENST00000610315.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700166584.hg.1 NM_001195278.1 Refseq
          Human Genome U133 Plus 2.0 Array 227318_at - -
          Human Genome U133 Plus 2.0 Array 227318_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227318_at7 - -
          Human Genome U133 Set 227318_at - -
          Human Genome U133 Set 227318_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 219235 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0