Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011737.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181161.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181161.hg.1 |
TC0500011737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181161.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
TC0500011737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181157.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
NR_130785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
TC0500011737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
ENST00000506265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
ENST00000514115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500181158.hg.1 |
NR_130785.1 |
Refseq |