Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03872223_m1
          See other SMC5-AS1 GE Assays ›
          Gene Symbol:
          SMC5-AS1
          Gene Name:
          SMC5 antisense RNA 1 (head to head)
          Gene Aliases:
          MAMDC2-AS2
          Chromosome Location:
          Chr.9: 70216059 - 70258874 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03872223_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SMC5-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_039990.1 2 - 3 491 69
          GenBank mRNA BC040840.1 - 2 - 3 491 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream MAM domain containing 2 MAMDC2 -2.2kb Anti-sense Chr. 9 - 70043519 - 70226972 on NCBI Build GRCh38
          Downstream structural maintenance of chromosomes 5 SMC5 SMC5L1 1.0kb Anti-sense Chr. 9 - 70258962 - 70354873 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900010362.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001199.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27059823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27059825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27059821 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118634 AJ223814.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118634 AK026934.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118634 BC106878.2 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118634 BC106879.2 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118634 NM_003162.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CGH Microarray 4x180K 58924 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 9454 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 797572 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 718693 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 585088 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 368859 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 211577 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 380897 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 935699 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18641805 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18641823 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18641823 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18641823 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18641805 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02102119 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 108853 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 157300 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 26083 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P26563871 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02102124 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02102138 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18641823 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02102119 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18641829 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18641809 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18641811 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18641805 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16854888 - -
          Agilent Human 1B Microarray G4111A A_32_P500625 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18641805 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P26563871 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18641823 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02102119 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09015336.hg.1 NR_039990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09015336.hg.1 TC09001199.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09015339.hg.1 NR_039990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09015339.hg.1 TC09001199.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09015337.hg.1 NR_039990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09015337.hg.1 TC09001199.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900170930.hg.1 NR_039990.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170930.hg.1 TC0900010362.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900170930.hg.1 NR_039990.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170936.hg.1 NR_039990.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170936.hg.1 TC0900010362.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900170936.hg.1 NR_039990.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170932.hg.1 ENST00000594708.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170932.hg.1 NR_039990.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170932.hg.1 TC0900010362.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900170932.hg.1 ENST00000594708.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170932.hg.1 NR_039990.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557693_PM_at - -
          Human Genome U133 Plus 2.0 Array 1557693_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557693_at3 - -
          Human Genome U95 Set 91359_r_at - -
          Human Genome U95 Set 91359_PM_r_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline