Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
ENST00000375466.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007308.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150305.hg.1 |
ENST00000332582.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00150305.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150305.hg.1 |
TC0X00007308.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00150305.hg.1 |
NM_018969.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00150305.hg.1 |
ENST00000332582.4 |
Ensembl |