Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03879019_s1
          See other STK32C GE Assays ›
          Gene Symbol:
          STK32C
          Gene Name:
          serine/threonine kinase 32C
          Gene Aliases:
          PKE, YANK3
          Chromosome Location:
          Chr.10: 132207482 - 132332559 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03879019_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: STK32C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318880.1 NP_001305809.1 2 531 75
          NM_001318883.1 NP_001305812.1 2 485 75
          GenBank mRNA AA782619.1 - 1 75
          BC042566.1 - 1 1452 75
          BF449072.1 - 1 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA864269.1 - 1 Chr. 10 - 132330293 - 132331650 [+] on Build GRCh38 75
          GenBankmRNA HY258372.1 - 1 Chr. 10 - 132330291 - 132331709 [+] on Build GRCh38 75
          GenBankmRNA AA398890.1 - 1 Chr. 10 - 132330291 - 132331652 [+] on Build GRCh38 75
          GenBankmRNA HY283502.1 - 1 Chr. 10 - 132330291 - 132330736 [+] on Build GRCh38 75
          GenBankmRNA BM678768.1 - 1 Chr. 10 - 132330291 - 132330661 [+] on Build GRCh38 75
          GenBankmRNA DN997246.1 - 2 500 Chr. 10 - 132330294 - 132331837 [-] on Build GRCh38 75
          GenBankmRNA AI867824.1 - 1 Chr. 10 - 132330289 - 132331552 [+] on Build GRCh38 75
          GenBankmRNA AI936830.1 - 1 Chr. 10 - 132330291 - 132331710 [+] on Build GRCh38 75
          GenBankmRNA HY282789.1 - 1 Chr. 10 - 132330291 - 132330784 [+] on Build GRCh38 75
          GenBankmRNA BM720428.1 - 1 266 Chr. 10 - 132330291 - 132330659 [-] on Build GRCh38 75
          GenBankmRNA AI368429.1 - 1 Chr. 10 - 132330296 - 132331595 [+] on Build GRCh38 75
          GenBankmRNA AI183653.1 - 1 Chr. 10 - 132330281 - 132331675 [+] on Build GRCh38 75
          GenBankmRNA AI024382.1 - 1 Chr. 10 - 132330291 - 132331590 [+] on Build GRCh38 75
          GenBankmRNA HY257743.1 - 1 Chr. 10 - 132330296 - 132330808 [+] on Build GRCh38 75
          GenBankmRNA DW426014.1 - 129 Chr. - - [-] on Build 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream leucine rich repeat containing 27 LRRC27 Anti-sense Chr. 10 - 132330063 - 132381506 on NCBI Build GRCh38
          Downstream PWWP domain containing 2B PWWP2B PWWP2,
          bA432J24.1,
          pp8607
          65.3kb Anti-sense Chr. 10 - 132397196 - 132417863 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-serine phosphorylation
          intracellular signal transduction
          protein serine/threonine kinase activity
          ATP binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000012612.hg.1 ENST00000368619.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012612.hg.1 ENST00000368619.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012612.hg.1 ENST00000456004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012612.hg.1 ENST00000456004.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012612.hg.1 ENST00000368619.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012612.hg.1 ENST00000368619.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000012612.hg.1 ENST00000456004.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012612.hg.1 ENST00000456004.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0