Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015343.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212365.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212365.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212365.hg.1 |
TC0200015343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200212365.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212365.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212364.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212364.hg.1 |
NR_046837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200212364.hg.1 |
TC0200015343.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200212364.hg.1 |
ENST00000428191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200212364.hg.1 |
NR_046837.1 |
Refseq |