Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03888814_s1
          See other LOC103611081 GE Assays ›
          Gene Symbol:
          LOC103611081 SNX19
          Gene Name:
          uncharacterized LOC103611081
          sorting nexin 19
          Gene Aliases:
          -
          CHET8
          Chromosome Location:
          Chr.11: 130866249 - 130870247 on Build GRCh38
          Chromosome Location:
          Chr.11: 130866254 - 130916488 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03888814_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC103611081
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_125382.1 1 3743 92
          GenBank mRNA AK124423.1 - 1 3743 92
          AL833360.1 - 1 1210 92
          Gene Symbol: SNX19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005271546.4 XP_005271603.1 10 15258 92
          XM_011542819.2 XP_011541121.1 13 15689 92
          XM_011542820.2 XP_011541122.1 13 15677 92
          XM_011542821.2 XP_011541123.1 12 15570 92
          XM_011542824.2 XP_011541126.1 13 14594 92
          XM_011542825.2 XP_011541127.1 13 13856 92
          XM_011542826.2 XP_011541128.1 13 13673 92
          XM_011542827.2 XP_011541129.1 12 13554 92
          XM_017017725.1 XP_016873214.1 10 15299 92
          XM_017017726.1 XP_016873215.1 11 13607 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI376428.1 - 1 Chr. 11 - 130866256 - 130866774 [+] on Build GRCh38 92
          GenBankmRNA AI142673.1 - 1 Chr. 11 - 130866255 - 130866705 [+] on Build GRCh38 92
          GenBankmRNA BF593032.1 - 1 Chr. 11 - 130866260 - 130866701 [+] on Build GRCh38 92
          GenBankmRNA AA826943.1 - 1 Chr. 11 - 130866253 - 130866574 [+] on Build GRCh38 92
          GenBankmRNA AI031541.1 - 1 Chr. 11 - 130866254 - 130866629 [+] on Build GRCh38 92
          GenBankmRNA DB296101.1 - 1 Chr. 11 - 130866257 - 130866795 [+] on Build GRCh38 92
          GenBankmRNA N56982.1 - 1 Chr. 11 - 130866250 - 130866724 [+] on Build GRCh38 92
          GenBankmRNA N34955.1 - 1 Chr. 11 - 130866252 - 130866697 [+] on Build GRCh38 92
          GenBankmRNA AW008845.1 - 1 Chr. 11 - 130866250 - 130866688 [+] on Build GRCh38 92
          GenBankmRNA AI439579.1 - 1 Chr. 11 - 130866261 - 130866715 [+] on Build GRCh38 92
          GenBankmRNA AW016840.1 - 1 Chr. 11 - 130866254 - 130866724 [+] on Build GRCh38 92
          GenBankmRNA AA805496.1 - 1 Chr. 11 - 130866253 - 130866705 [+] on Build GRCh38 92
          GenBankmRNA BP396117.1 - 1 260 Chr. 11 - 130866265 - 130866763 [-] on Build GRCh38 92
          GenBankmRNA AI024416.1 - 1 Chr. 11 - 130866252 - 130866716 [+] on Build GRCh38 92
          GenBankmRNA AI126709.1 - 1 Chr. 11 - 130866250 - 130866646 [+] on Build GRCh38 92
          GenBankmRNA AI814777.1 - 1 Chr. 11 - 130866257 - 130866738 [+] on Build GRCh38 92
          GenBankmRNA BX115669.1 - 1 588 Chr. 11 - 130866389 - 130867093 [-] on Build GRCh38 92
          GenBankmRNA BF447990.1 - 1 Chr. 11 - 130866250 - 130866549 [+] on Build GRCh38 92
          GenBankmRNA BM979366.1 - 1 Chr. 11 - 130866254 - 130866971 [+] on Build GRCh38 92
          GenBankmRNA AI140391.1 - 1 Chr. 11 - 130866250 - 130866646 [+] on Build GRCh38 92
          GenBankmRNA AW001058.1 - 1 Chr. 11 - 130866261 - 130866766 [+] on Build GRCh38 92
          GenBankmRNA AI332857.1 - 1 Chr. 11 - 130866263 - 130866636 [+] on Build GRCh38 92
          GenBankmRNA AI246249.1 - 1 Chr. 11 - 130866259 - 130866564 [+] on Build GRCh38 92
          GenBankmRNA R37438.1 - Chr. - - [-] on Build 92
          GenBankmRNA BU620819.1 - 1 Chr. 11 - 130866254 - 130866911 [+] on Build GRCh38 92
          GenBankmRNA BE856778.1 - 1 Chr. 11 - 130866250 - 130866765 [+] on Build GRCh38 92
          GenBankmRNA AI088181.1 - 1 Chr. 11 - 130866254 - 130866621 [+] on Build GRCh38 92
          GenBankmRNA BM974069.1 - 1 Chr. 11 - 130866250 - 130867033 [+] on Build GRCh38 92
          GenBankmRNA AI204416.1 - 1 Chr. 11 - 130866255 - 130866544 [+] on Build GRCh38 92
          GenBankmRNA CA388895.1 - 1 Chr. 11 - 130866299 - 130866763 [+] on Build GRCh38 92
          GenBankmRNA CA311671.1 - 1 Chr. 11 - 130866250 - 130866911 [+] on Build GRCh38 92
          GenBankmRNA AW977676.1 - 1 Chr. 11 - 130866254 - 130866750 [+] on Build GRCh38 92
          GenBankmRNA BF221867.1 - 1 Chr. 11 - 130866256 - 130866745 [+] on Build GRCh38 92
          GenBankmRNA AW182612.1 - 1 Chr. 11 - 130866256 - 130866652 [+] on Build GRCh38 92
          GenBankmRNA DB352376.1 - 1 Chr. 11 - 130866257 - 130866809 [+] on Build GRCh38 92
          GenBankmRNA DB315350.1 - 1 Chr. 11 - 130866251 - 130866799 [+] on Build GRCh38 92
          GenBankmRNA AI671383.1 - 1 Chr. 11 - 130866237 - 130866626 [+] on Build GRCh38 92
          GenBankmRNA AA629392.1 - 1 Chr. 11 - 130866255 - 130866618 [+] on Build GRCh38 92
          GenBankmRNA BU632358.1 - 1 Chr. 11 - 130866254 - 130866799 [+] on Build GRCh38 92
          GenBankmRNA AA256267.1 - 1 Chr. 11 - 130866249 - 130866616 [+] on Build GRCh38 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100507431 LOC100507431 -4.1kb Sense Chr. 11 - 130844193 - 130862110 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100012852.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          chondrocyte differentiation
          exocytosis
          insulin secretion
          dense core granule maturation
          protein binding
          phosphatidylinositol-3-phosphate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533434.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533434.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533434.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533434.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533434.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533434.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533434.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533434.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 ENST00000533812.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Clariom D Assay TC1100012852.hg.1 NR_125383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198568.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198568.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198568.hg.1 TC1100012852.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100198568.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198568.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198567.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198567.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198567.hg.1 TC1100012852.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100198567.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198567.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198572.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198572.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198572.hg.1 TC1100012852.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100198572.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198572.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198570.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198570.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198570.hg.1 TC1100012852.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100198570.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198570.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198571.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198571.hg.1 NR_125382.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100198571.hg.1 TC1100012852.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100198571.hg.1 ENST00000525716.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100198571.hg.1 NR_125382.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc