Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03899466_s1
          See other LZIC GE Assays ›
          Gene Symbol:
          LZIC
          Gene Name:
          leucine zipper and CTNNBIP1 domain containing
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 9922113 - 9943427 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03899466_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LZIC
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001316973.1 NP_001303902.1 8 1857 89
          NM_001316974.1 NP_001303903.1 8 1800 89
          NM_001316976.1 NP_001303905.1 6 1679 89
          NM_032368.4 NP_115744.2 8 1957 89
          XM_005263509.2 XP_005263566.1 6 1674 89
          XM_017002546.1 XP_016858035.1 8 1703 89
          XM_017002547.1 XP_016858036.1 8 1723 89
          GenBank mRNA AK025913.1 - 1 1054 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI804235.1 - 1 Chr. 1 - 9929281 - 9929731 [+] on Build GRCh38 89
          GenBankmRNA AI346534.1 - 1 Chr. 1 - 9929281 - 9929740 [+] on Build GRCh38 89
          GenBankmRNA AW770988.1 - 1 Chr. 1 - 9929284 - 9929755 [+] on Build GRCh38 89
          GenBankmRNA CK819912.1 - 1 Chr. 1 - 9928937 - 9929475 [+] on Build GRCh38 89
          GenBankmRNA AI741164.1 - 1 Chr. 1 - 9929282 - 9929757 [+] on Build GRCh38 89
          GenBankmRNA AW013858.1 - 1 Chr. 1 - 9929281 - 9929762 [+] on Build GRCh38 89
          GenBankmRNA AI589613.1 - 1 Chr. 1 - 9929281 - 9929741 [+] on Build GRCh38 89
          GenBankmRNA BE219717.1 - 1 Chr. 1 - 9929282 - 9929791 [+] on Build GRCh38 89
          GenBankmRNA BM141836.1 - 1 Chr. 1 - 9928937 - 9929473 [+] on Build GRCh38 89
          GenBankmRNA DB303824.1 - 1 Chr. 1 - 9928938 - 9929474 [+] on Build GRCh38 89
          GenBankmRNA AI350775.1 - 1 Chr. 1 - 9929280 - 9929603 [+] on Build GRCh38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream catenin beta interacting protein 1 CTNNBIP1 ICAT -18.7kb Sense Chr. 1 - 9848276 - 9910322 on NCBI Build GRCh38
          Downstream nicotinamide nucleotide adenylyltransferase 1 NMNAT1 LCA9,
          NMNAT,
          PNAT1
          12.5kb Anti-sense Chr. 1 - 9942923 - 9996739 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LZIC

          Gene Ontology Categories:

          Function(s) Process(es)

          response to ionizing radiation
          beta-catenin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000471853.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000471853.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000471853.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000471853.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377213.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000377223.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000400903.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000471853.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000471853.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000471853.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000471853.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 ENST00000488540.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012828.hg.1 NM_032368.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0