Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03899538_s1
          See other ELK3 GE Assays ›
          Gene Symbol:
          ELK3
          Gene Name:
          ELK3, ETS transcription factor
          Gene Aliases:
          ERP, NET, SAP-2, SAP2
          Chromosome Location:
          Chr.12: 96194382 - 96269835 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03899538_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ELK3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001303511.1 NP_001290440.1 3 2191 106
          NM_005230.3 NP_005221.2 5 3428 106
          XM_006719275.3 XP_006719338.1 6 3525 106
          GenBank mRNA AK026078.1 - 1 1346 106
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA R91287.1 - Chr. - - [-] on Build 106
          GenBankmRNA AW368149.1 - 1 Chr. 12 - 96268565 - 96269093 [-] on Build GRCh38 106
          GenBankmRNA H53750.1 - 1 312 Chr. 12 - 96268747 - 96269139 [+] on Build GRCh38 106
          GenBankmRNA CB120627.1 - 1 338 Chr. 12 - 96268721 - 96269173 [+] on Build GRCh38 106
          GenBankmRNA AW405395.1 - 1 86 Chr. 12 - 96268980 - 96269349 [+] on Build GRCh38 106
          GenBankmRNA AA029744.1 - 1 301 Chr. 12 - 96268759 - 96269434 [+] on Build GRCh38 106
          GenBankmRNA BX110517.1 - 1 479 Chr. 12 - 96268580 - 96269425 [+] on Build GRCh38 106
          GenBankmRNA BP419864.1 - 1 384 Chr. 12 - 96268675 - 96269123 [+] on Build GRCh38 106
          GenBankmRNA AW372522.1 - 1 Chr. 12 - 96268790 - 96269409 [-] on Build GRCh38 106
          GenBankmRNA H97146.1 - 1 Chr. 12 - 96268887 - 96269181 [-] on Build GRCh38 106
          GenBankmRNA BX436694.2 - 1 Chr. 12 - 96268847 - 96269795 [-] on Build GRCh38 106
          GenBankmRNA BF951798.1 - 1 249 Chr. 12 - 96268822 - 96269383 [+] on Build GRCh38 106
          GenBankmRNA CN361136.1 - 1 403 Chr. 12 - 96268658 - 96269426 [+] on Build GRCh38 106
          GenBankmRNA CN361137.1 - 1 495 Chr. 12 - 96268565 - 96269330 [+] on Build GRCh38 106
          GenBankmRNA AA299256.1 - 1 99 Chr. 12 - 96268960 - 96269223 [+] on Build GRCh38 106
          GenBankmRNA BG250780.1 - 436 Chr. - - [-] on Build 106
          GenBankmRNA CB119521.1 - 1 325 Chr. 12 - 96268733 - 96269175 [+] on Build GRCh38 106
          GenBankmRNA BM460796.1 - 504 Chr. - - [-] on Build 106
          GenBankmRNA BF951795.1 - 1 246 Chr. 12 - 96268829 - 96269374 [+] on Build GRCh38 106
          GenBankmRNA AW372523.1 - 1 Chr. 12 - 96268807 - 96269421 [-] on Build GRCh38 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream leukotriene A4 hydrolase LTA4H -224.2kb Anti-sense Chr. 12 - 96000753 - 96043520 on NCBI Build GRCh38
          Downstream cyclin dependent kinase 17 CDK17 PCTAIRE2,
          PCTK2
          8.4kb Anti-sense Chr. 12 - 96278261 - 96400588 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ELK3

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          transcription by RNA polymerase II
          signal transduction
          cell differentiation
          wound healing
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity
          transcription corepressor activity
          protein binding
          purine-rich negative regulatory element binding

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 164526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0