Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03902605_s1
          See other GNAQ GE Assays ›
          Gene Symbol:
          GNAQ
          Gene Name:
          G protein subunit alpha q
          Gene Aliases:
          CMC1, G-ALPHA-q, GAQ, SWS
          Chromosome Location:
          Chr.9: 77716274 - 78031449 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03902605_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GNAQ
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002072.4 NP_002063.2 7 6024 71
          GenBank mRNA AK055619.1 - 1 1851 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA031622.1 - 1 Chr. 9 - 77716274 - 77716722 [+] on Build GRCh38 71
          GenBankmRNA AU253465.1 - 1 Chr. 9 - 77716284 - 77716976 [+] on Build GRCh38 71
          GenBankmRNA DB319874.1 - 1 Chr. 9 - 77716276 - 77716809 [+] on Build GRCh38 71
          GenBankmRNA HY319685.1 - 1 Chr. 9 - 77716274 - 77716685 [+] on Build GRCh38 71
          GenBankmRNA BQ010386.1 - 1 Chr. 9 - 77716274 - 77716878 [+] on Build GRCh38 71
          GenBankmRNA AW872409.1 - 1 Chr. 9 - 77716261 - 77716678 [+] on Build GRCh38 71
          GenBankmRNA BI757085.1 - 1 501 Chr. 9 - 77716211 - 77717084 [-] on Build GRCh38 71
          GenBankmRNA BM663324.1 - 1 Chr. 9 - 77716206 - 77716647 [+] on Build GRCh38 71
          GenBankmRNA AI608984.1 - 1 Chr. 9 - 77716274 - 77716737 [+] on Build GRCh38 71
          GenBankmRNA AI342963.1 - 1 Chr. 9 - 77716275 - 77716632 [+] on Build GRCh38 71
          GenBankmRNA AA149174.1 - 1 Chr. 9 - 77716276 - 77716681 [+] on Build GRCh38 71
          GenBankmRNA AA031703.1 - 1 205 Chr. 9 - 77716278 - 77716797 [-] on Build GRCh38 71
          GenBankmRNA BM667065.1 - 1 Chr. 9 - 77716274 - 77716764 [+] on Build GRCh38 71
          GenBankmRNA AW665760.1 - 1 Chr. 9 - 77716274 - 77716772 [+] on Build GRCh38 71
          GenBankmRNA CB243775.1 - 1 Chr. 9 - 77716274 - 77717066 [+] on Build GRCh38 71
          GenBankmRNA AA044282.1 - 1 Chr. 9 - 77716276 - 77716708 [+] on Build GRCh38 71
          GenBankmRNA AW673507.1 - 1 Chr. 9 - 77716209 - 77716697 [+] on Build GRCh38 71
          GenBankmRNA AW901974.1 - 1 329 Chr. 9 - 77716509 - 77716920 [-] on Build GRCh38 71
          GenBankmRNA BM978904.1 - 1 Chr. 9 - 77716095 - 77716675 [+] on Build GRCh38 71
          GenBankmRNA AI800762.1 - 1 Chr. 9 - 77716276 - 77716749 [+] on Build GRCh38 71
          GenBankmRNA BM479765.1 - 1 171 Chr. 9 - 77716185 - 77716763 [-] on Build GRCh38 71
          GenBankmRNA AI707842.1 - 1 Chr. 9 - 77716274 - 77716634 [+] on Build GRCh38 71
          GenBankmRNA BE178532.1 - 1 Chr. 9 - 77716526 - 77716899 [+] on Build GRCh38 71
          GenBankmRNA AA001858.1 - 1 Chr. 9 - 77716274 - 77716722 [+] on Build GRCh38 71
          GenBankmRNA BE702186.1 - 1 332 Chr. 9 - 77716558 - 77716920 [-] on Build GRCh38 71
          GenBankmRNA AA026157.1 - 1 205 Chr. 9 - 77716378 - 77716797 [-] on Build GRCh38 71
          GenBankmRNA BM690633.1 - 62 Chr. - - [-] on Build 71
          GenBankmRNA BQ130130.1 - 1 Chr. 9 - 77716091 - 77716697 [+] on Build GRCh38 71
          GenBankmRNA BE856684.1 - 1 Chr. 9 - 77716274 - 77716806 [+] on Build GRCh38 71
          GenBankmRNA AA136764.1 - 1 244 Chr. 9 - 77716364 - 77716836 [-] on Build GRCh38 71
          GenBankmRNA AA659915.1 - 1 Chr. 9 - 77716275 - 77716655 [+] on Build GRCh38 71
          GenBankmRNA AI375034.1 - 1 Chr. 9 - 77716274 - 77716710 [+] on Build GRCh38 71
          GenBankmRNA BM979946.1 - 1 Chr. 9 - 77716274 - 77716980 [+] on Build GRCh38 71
          GenBankmRNA AA149173.1 - 1 215 Chr. 9 - 77716378 - 77716801 [-] on Build GRCh38 71
          GenBankmRNA BF109469.1 - 1 Chr. 9 - 77716276 - 77716721 [+] on Build GRCh38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream G protein subunit alpha 14 GNA14 -68.0kb Sense Chr. 9 - 77423079 - 77648316 on NCBI Build GRCh38
          Downstream centrosomal protein 78 CEP78 C9orf81,
          IP63
          517.6kb Anti-sense Chr. 9 - 78236062 - 78278431 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900010485.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GNAQ

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          action potential
          negative regulation of protein kinase activity
          adenylate cyclase-activating G protein-coupled receptor signaling pathway
          activation of phospholipase C activity
          G protein-coupled acetylcholine receptor signaling pathway
          glutamate receptor signaling pathway
          heart development
          blood coagulation
          phototransduction, visible light
          entrainment of circadian clock
          post-embryonic development
          neuron remodeling
          forebrain neuron development
          platelet activation
          maternal behavior
          embryonic digit morphogenesis
          positive regulation of GTPase activity
          regulation of melanocyte differentiation
          developmental pigmentation
          protein stabilization
          phospholipase C-activating dopamine receptor signaling pathway
          regulation of canonical Wnt signaling pathway
          GTPase activity
          GTPase activator activity
          protein binding
          GTP binding
          G-protein beta/gamma-subunit complex binding
          type 2A serotonin receptor binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000411677.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000411677.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000411677.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 ENST00000411677.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000411677.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000411677.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000411677.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 ENST00000411677.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay TC0900010485.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900171724.hg.1 ENST00000286548.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900171724.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900171724.hg.1 TC0900010485.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900171724.hg.1 NM_002072.4 Refseq
          Affymetrix Human Clariom D Assay PSR0900171724.hg.1 ENST00000286548.8 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 171172 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0