Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03902752_s1
          See other CLVS2 GE Assays ›
          Gene Symbol:
          CLVS2
          Gene Name:
          clavesin 2
          Gene Aliases:
          C6orf212, C6orf213, RLBP1L2, bA160A10.4
          Chromosome Location:
          Chr.6: 122993693 - 123081282 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03902752_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CLVS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010852.3 NP_001010852.2 6 10702 100
          XM_017010266.1 XP_016865755.1 6 12404 100
          GenBank mRNA AK055705.1 - 1 1846 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY151725.1 - 1 422 Chr. 6 - 123071724 - 123072187 [+] on Build GRCh38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream sphingomyelin phosphodiesterase acid like 3A SMPDL3A ASM3A,
          ASML3a,
          yR36GH4.1
          -260.6kb Sense Chr. 6 - 122787180 - 122809720 on NCBI Build GRCh38
          Downstream triadin TRDN CPVT5,
          TDN,
          TRISK
          143.4kb Anti-sense Chr. 6 - 123216339 - 123637093 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600009336.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CLVS2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          lysosome organization
          transporter activity
          lipid binding
          phosphatidylinositol-3,5-bisphosphate binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163953.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163953.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163953.hg.1 TC0600009336.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600163953.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163953.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163957.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163957.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163957.hg.1 TC0600009336.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600163957.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163957.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163954.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163954.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163954.hg.1 TC0600009336.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600163954.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163954.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163955.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163955.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163955.hg.1 TC0600009336.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600163955.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163955.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 ENST00000368438.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay TC0600009336.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163956.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163956.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163956.hg.1 TC0600009336.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600163956.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163956.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163952.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163952.hg.1 NM_001010852.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600163952.hg.1 TC0600009336.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600163952.hg.1 ENST00000275162.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600163952.hg.1 NM_001010852.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 126802 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0