Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011547.hg.1 |
ENST00000432247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000356684.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000356684.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000356684.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000356684.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000424044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000424044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
NR_027285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
NR_027285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
NR_027285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
NR_027285.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
NR_027286.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
NR_027286.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017262.hg.1 |
NR_027286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011547.hg.1 |
ENST00000432247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000356684.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000356684.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000356684.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000356684.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000424044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000424044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
ENST00000426161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
NR_027285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
NR_027285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
NR_027285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
NR_027285.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
NR_027286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
NR_027286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017262.hg.1 |
NR_027286.1 |
Refseq |