Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03918355_s1
          See other LOC100505782 GE Assays ›
          Gene Symbol:
          LOC100505782
          Gene Name:
          uncharacterized LOC100505782
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 41402416 - 41412588 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03918355_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100505782
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040111.1 3 2053 104
          GenBank mRNA BC039320.1 - 3 2053 104

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream keratin 31 KRT31 HA1,
          Ha-1,
          KRTHA1,
          hHa1
          -14.8kb Anti-sense Chr. 17 - 41393724 - 41397592 on NCBI Build GRCh38
          Downstream keratin 37 KRT37 HA7,
          K37,
          KRTHA7
          8.0kb Anti-sense Chr. 17 - 41420557 - 41424570 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007834.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000511.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10322489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30894507 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 174661 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 729024 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 520964 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 218928 BC017907.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 218928 BC033168.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 218928 BC103755.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 218928 NM_057179.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 955372 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 104716 M11353.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 105504 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03245119 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P40817537 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03245119 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 29905 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 33095 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03245110 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03245119 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03245123 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20647431 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20647464 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40817536 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20647443 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40817537 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P20863904 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03245134 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03245130 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03245130 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P40817537 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17006110.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006110.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006110.hg.1 NR_040111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006110.hg.1 TC17000511.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17006113.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006113.hg.1 NR_040111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006113.hg.1 TC17000511.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 NR_040111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 NR_040111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007834.hg.1 NR_040111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006111.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006111.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17006111.hg.1 NR_040111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17006111.hg.1 TC17000511.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700156959.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156959.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156959.hg.1 NR_040111.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700156959.hg.1 TC1700007834.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700156959.hg.1 NR_040111.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700156959.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156959.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 NR_040111.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 TC1700007834.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156956.hg.1 NR_040111.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 NR_040111.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 TC1700007834.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700156957.hg.1 NR_040111.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000430006.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 ENST00000432258.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 NR_040111.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 NR_040111.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007834.hg.1 NR_040111.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557068_PM_at - -
          Human Genome U133 Plus 2.0 Array 1557068_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557068_at9 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009514 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009514 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 151325 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0