Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03918716_s1
          See other GS1-24F4.2 GE Assays ›
          Gene Symbol:
          GS1-24F4.2
          Gene Name:
          uncharacterized LOC100652791
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 6835554 - 6842454 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03918716_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GS1-24F4.2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045217.1 3 950 108
          GenBank mRNA BC043547.1 - 3 950 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI806287.1 - 1 Chr. 8 - 6842053 - 6842472 [-] on Build GRCh38 108
          GenBankmRNA CD557887.1 - 1 615 Chr. 8 - 6841693 - 6842571 [+] on Build GRCh38 108
          GenBankmRNA AI688579.1 - 1 Chr. 8 - 6841941 - 6842445 [-] on Build GRCh38 108
          GenBankmRNA AI240483.1 - 1 Chr. 8 - 6842101 - 6842472 [-] on Build GRCh38 108

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream XK related 5 XKR5 HARL2754,
          UNQ2754,
          XRG5A,
          XRG5BM
          -6.0kb Anti-sense Chr. 8 - 6808517 - 6835644 on NCBI Build GRCh38
          Downstream defensin beta 1 DEFB1 BD1,
          DEFB-1,
          DEFB101,
          HBD1
          28.1kb Anti-sense Chr. 8 - 6870575 - 6878007 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800006573.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000024.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05940720 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05940721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05940722 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05940724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26214186 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 58213 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 611091 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 328013 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P145769 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P145764 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 37099 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 40544 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18204909 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P145769 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P145764 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18204909 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18204909 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P145769 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P145764 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 18314 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 10772 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18204909 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18204906 - -
          Agilent Human 1B Microarray G4111A A_32_P145764 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18204909 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08000363.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08000363.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08000363.hg.1 TC08000024.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08000364.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08000364.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08000364.hg.1 TC08000024.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08000365.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08000365.hg.1 NR_045217.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08000365.hg.1 TC08000024.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0800144804.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144804.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144804.hg.1 TC0800006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800144804.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144804.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144807.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144807.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144807.hg.1 TC0800006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800144807.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144807.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144805.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144805.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144805.hg.1 TC0800006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800144805.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144805.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144809.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144809.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144809.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144809.hg.1 TC0800006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800144809.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144809.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144809.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144810.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144810.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144810.hg.1 TC0800006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800144810.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144810.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144811.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144811.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144811.hg.1 TC0800006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800144811.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144811.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144808.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144808.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800144808.hg.1 TC0800006573.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800144808.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800144808.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000500823.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 ENST00000531701.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 NR_045217.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006573.hg.1 NR_045217.1 Refseq
          Human Genome U133 Plus 2.0 Array 1559678_s_at - -
          Human Genome U133 Plus 2.0 Array 1559678_PM_s_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559678_s_at4 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline