Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03921574_s1
          See other ERV9-1 GE Assays ›
          Gene Symbol:
          ERV9-1
          Gene Name:
          endogenous retrovirus group 9 member 1
          Gene Aliases:
          pTR2
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03921574_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ERV9-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA X15675.1 - 2 1751 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AU148611.1 - 1 Chr. 11 - 67626881 - 67627389 [-] on Build GRCh38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream double C2 domain gamma pseudogene DOC2GP -10.4kb Anti-sense Chr. 11 - 67613524 - 67615664 on NCBI Build GRCh38
          Downstream nudix hydrolase 8 NUDT8 545.0 Anti-sense Chr. 11 - 67621421 - 67630116 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 540762 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 757631 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 405577 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P32406 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15139 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P32406 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19270133 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P32406 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19270133 - -
          Agilent Human 1B Microarray G4111A A_32_P32407 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19270133 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400007158.hg.1 ENST00000503299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007158.hg.1 ENST00000503299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007158.hg.1 ENST00000503299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011345.hg.1 ENST00000533311.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011345.hg.1 ENST00000533311.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011345.hg.1 ENST00000533311.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011345.hg.1 ENST00000533311.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000422780.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000514568.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006733.hg.1 ENST00000540299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007158.hg.1 ENST00000503299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007158.hg.1 ENST00000503299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007158.hg.1 ENST00000503299.1 Ensembl
          Human Genome U95 Set 35585_PM_at - -
          Human Genome U95 Set 35585_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 477963 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d58dc5f8-fsfvr:80/100.66.134.75:80.
          git-commit: 38b8a393e477ae4d9d89cc5f7a29948d21ae29e2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc