Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03922839_s1
          See other MAN1A1 GE Assays ›
          Gene Symbol:
          MAN1A1
          Gene Name:
          mannosidase alpha class 1A member 1
          Gene Aliases:
          HUMM3, HUMM9, MAN9
          Chromosome Location:
          Chr.6: 119177201 - 119350619 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03922839_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAN1A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005907.3 NP_005898.2 13 4229 92
          XM_005266986.4 XP_005267043.1 13 4085 92
          XM_011535833.2 XP_011534135.1 12 3281 92
          GenBank mRNA AK024588.1 - 1 453 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX387461.2 - 1 Chr. 6 - 119177228 - 119178213 [+] on Build GRCh38 92
          GenBankmRNA AV759454.1 - 1 346 Chr. 6 - 119177790 - 119178323 [-] on Build GRCh38 92
          GenBankmRNA BG402600.1 - 1 348 Chr. 6 - 119177671 - 119178280 [-] on Build GRCh38 92
          GenBankmRNA BG616781.1 - 1 431 Chr. 6 - 119177718 - 119178429 [-] on Build GRCh38 92
          GenBankmRNA BF665216.1 - 450 Chr. - - [-] on Build 92
          GenBankmRNA BQ942728.1 - 1 434 Chr. 6 - 119177616 - 119178432 [-] on Build GRCh38 92
          GenBankmRNA BM313549.1 - 1 72 Chr. 6 - 119177534 - 119178054 [-] on Build GRCh38 92
          GenBankmRNA BG565453.1 - 1 61 Chr. 6 - 119177427 - 119178059 [-] on Build GRCh38 92
          GenBankmRNA AA262830.1 - 1 97 Chr. 6 - 119177724 - 119178095 [-] on Build GRCh38 92
          GenBankmRNA BG623671.1 - 1 359 Chr. 6 - 119177622 - 119178357 [-] on Build GRCh38 92
          GenBankmRNA AW965136.1 - 1 228 Chr. 6 - 119177581 - 119178226 [-] on Build GRCh38 92
          GenBankmRNA BM128096.1 - 1 215 Chr. 6 - 119177685 - 119178213 [-] on Build GRCh38 92
          GenBankmRNA AI114437.1 - 1 256 Chr. 6 - 119177623 - 119178213 [-] on Build GRCh38 92
          GenBankmRNA BX102630.1 - 1 555 Chr. 6 - 119177854 - 119178553 [-] on Build GRCh38 92
          GenBankmRNA BG548683.1 - 1 427 Chr. 6 - 119177662 - 119178425 [-] on Build GRCh38 92
          GenBankmRNA AV714881.1 - 2 531 Chr. 6 - 119177940 - 119302104 [-] on Build GRCh38 92
          GenBankmRNA HY255826.1 - 1 Chr. 6 - 119177652 - 119178124 [+] on Build GRCh38 92
          GenBankmRNA AV709183.1 - 1 341 Chr. 6 - 119177940 - 119178339 [-] on Build GRCh38 92
          GenBankmRNA BP382117.1 - 1 453 Chr. 6 - 119177838 - 119178451 [-] on Build GRCh38 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream family with sequence similarity 184 member A FAM184A C6orf60 -28.0kb Sense Chr. 6 - 118959829 - 119149193 on NCBI Build GRCh38
          Downstream uncharacterized LOC285762 LOC285762 274.1kb Sense Chr. 6 - 119452547 - 119491302 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAN1A1

          Gene Ontology Categories:

          Function(s) Process(es)

          N-glycan processing
          Golgi apparatus mannose trimming
          mannosyl-oligosaccharide 1,2-alpha-mannosidase activity
          calcium ion binding
          mannosidase activity

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 158279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0