Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03927348_s1
          See other LOC100288238 GE Assays ›
          Gene Symbol:
          LOC100288238
          Gene Name:
          uncharacterized LOC100288238
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03927348_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100288238
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC043439.2 - 1 449 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI561342.1 - 1 453 Chr. 9 - 21395619 - 21396346 [-] on Build GRCh38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream interferon alpha 2 IFNA2 IFN-alphaA,
          IFNA,
          IFNA2B,
          INFA2
          -9.5kb Sense Chr. 9 - 21384255 - 21385397 on NCBI Build GRCh38
          Downstream interferon alpha 8 IFNA8 IFN-alphaB 12.8kb Anti-sense Chr. 9 - 21409147 - 21410185 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638373 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638392 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638375 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638391 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638395 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638394 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638381 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06638393 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926562 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26926576 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1562880_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0900009708.hg.1 ENST00000569618.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009708.hg.1 ENST00000569618.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1562880_PM_at - -
          Human Genome U133 Plus 2.0 Array 1562880_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562880_at7 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1895504 BC043439.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1895504 BC043439.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1895504 BC043439.2 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 3290743 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_115205 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1895504 BC043439.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1895504 BC043439.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1895504 BC043439.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0