Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001718.hg.1 |
ENST00000377791.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001718.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001718.hg.1 |
TC06000167.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011231.hg.1 |
ENST00000627472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001720.hg.1 |
ENST00000314088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001720.hg.1 |
ENST00000377791.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001720.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001720.hg.1 |
TC06000167.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011132.hg.1 |
ENST00000629747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011231.hg.1 |
ENST00000627472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
TC0600007270.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149316.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
TC0600007270.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149317.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011132.hg.1 |
ENST00000629747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
ENST00000602637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007270.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149314.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149314.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149314.hg.1 |
NM_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149314.hg.1 |
TC0600007270.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149314.hg.1 |
ENST00000314088.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149314.hg.1 |
ENST00000377791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149314.hg.1 |
NM_003512.3 |
Refseq |