Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04188074_m1
          See other MRPL18 GE Assays ›
          Gene Symbol:
          MRPL18
          Gene Name:
          mitochondrial ribosomal protein L18
          Gene Aliases:
          HSPC071, L18mt, MRP-L18
          Chromosome Location:
          Chr.6: 159789812 - 159798429 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04188074_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MRPL18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318817.1 NP_001305746.1 2 - 3 380 76
          NM_014161.4 NP_054880.2 2 - 3 380 76
          NR_134860.1 3 - 4 500 76
          NR_134861.1 2 - 3 457 76
          NR_134862.1 2 - 3 380 76
          NR_134864.1 2 - 3 263 76
          NR_134865.1 2 - 3 380 76
          NR_134866.1 2 - 3 380 76
          GenBank mRNA AF161556.1 - 2 - 3 358 76
          AK311391.1 - 3 - 4 377 76
          AL136633.1 - 2 - 3 330 76
          AV756740.1 - 2 - 3 315 76
          BC001623.1 - 2 - 3 351 3357333 76
          BF797811.1 - 2 - 3 263 76
          BG548185.1 - 2 - 3 359 76
          BM924328.1 - 2 - 3 473 76
          BU601065.1 - 2 - 3 321 76
          BU932963.1 - 2 - 3 369 76
          BU943173.1 - 383 76
          CA777760.1 - 2 - 3 262 76
          CR533491.1 - 2 - 3 238 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600010060.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001148.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MRPL18

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          rRNA import into mitochondrion
          mitochondrial translational elongation
          mitochondrial translational termination
          structural constituent of ribosome
          5S rRNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06013664.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013664.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013664.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06013664.hg.1 TC06001148.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06013659.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013659.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06013659.hg.1 TC06001148.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06013663.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013663.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013663.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06013663.hg.1 TC06001148.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06013660.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013660.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013660.hg.1 NM_014161.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06013660.hg.1 TC06001148.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600168723.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168723.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168723.hg.1 TC0600010060.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600168723.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168723.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168725.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168725.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168725.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168725.hg.1 TC0600010060.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600168725.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168725.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168725.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 TC0600010060.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168729.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010060.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168724.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168724.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168724.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168724.hg.1 TC0600010060.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600168724.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168724.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168724.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 NM_014161.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 TC0600010060.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 ENST00000367034.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 ENST00000476826.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 ENST00000479638.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 ENST00000480842.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600168728.hg.1 NM_014161.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 197989 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0