Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04192465_gH
          See other HLA-DRB1 GE Assays ›
          Gene Symbol:
          HLA-DRB1
          Gene Name:
          major histocompatibility complex, class II, DR beta 1
          Gene Aliases:
          DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
          Chromosome Location:
          Chr.6: 32578769 - 32589836 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04192465_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HLA-DRB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002124.3 NP_002115.2 4 - 5 858 99
          GenBank mRNA AK289463.1 - 4 - 5 827 99
          AK291987.1 - 4 - 5 858 99
          BC033827.1 - 4 - 5 807 5216243 99
          BC108922.1 - 4 - 5 819 40023604 99
          M20138.1 - 4 - 5 766 99
          M20430.1 - 4 - 5 807 99
          M20504.1 - 5 - 6 799 99
          M28583.1 - 5 - 6 803 99
          M28584.1 - 4 - 5 735 99
          M33600.1 - 4 - 5 834 99
          U56640.1 - 3 - 4 621 99
          X00700.1 - 4 - 5 768 99
          X03069.1 - 4 - 5 826 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014273.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06004123.hg.1, TC6_cox_hap2000193.hg.1, TC6_qbl_hap6000182.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HLA-DRB1

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of exogenous peptide antigen via MHC class II
          T cell costimulation
          T cell receptor signaling pathway
          interferon-gamma-mediated signaling pathway
          MHC class II protein complex binding
          polysaccharide binding
          peptide antigen binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06018659.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06018659.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018659.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018659.hg.1 TC06004123.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003800.hg.1 TC6_cox_hap2000193.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003800.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003800.hg.1 NM_022555.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003634.hg.1 TC6_qbl_hap6000182.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003634.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018654.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06018654.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018654.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018654.hg.1 TC06004123.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06018649.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06018649.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018649.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018649.hg.1 TC06004123.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003801.hg.1 TC6_cox_hap2000193.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003801.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003801.hg.1 NM_022555.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003658.hg.1 TC6_qbl_hap6000182.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003658.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018647.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06018647.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018647.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018647.hg.1 TC06004123.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003633.hg.1 TC6_qbl_hap6000182.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003633.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003662.hg.1 TC6_qbl_hap6000182.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003662.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003662.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018662.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06018662.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018662.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06018662.hg.1 TC06004123.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003821.hg.1 TC6_cox_hap2000193.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003821.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003821.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003825.hg.1 TC6_cox_hap2000193.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003825.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003828.hg.1 TC6_cox_hap2000193.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003828.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003828.hg.1 NM_002124.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003654.hg.1 TC6_qbl_hap6000182.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003654.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003654.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600201471.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600201471.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600201471.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600201471.hg.1 TC0600014273.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600201471.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600201471.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600201471.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600201464.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600201464.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600201464.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600201464.hg.1 TC0600014273.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600201464.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600201464.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600201464.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600201465.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600201465.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600201465.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600201465.hg.1 TC0600014273.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600201465.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600201465.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600201465.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600201466.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600201466.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600201466.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600201466.hg.1 TC0600014273.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600201466.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600201466.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600201466.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 ENST00000360004.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_001243965.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014273.hg.1 NM_002124.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 140522 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0