Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027820.hg.1 |
ENST00000367360.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027820.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027820.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027820.hg.1 |
TC06002220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027822.hg.1 |
ENST00000367360.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027822.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027822.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027822.hg.1 |
TC06002220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027821.hg.1 |
ENST00000367360.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027821.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027821.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027821.hg.1 |
TC06002220.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0600193508.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193508.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193508.hg.1 |
TC0600013537.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193508.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193508.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
TC0600013537.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193506.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193507.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193507.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193507.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193507.hg.1 |
TC0600013537.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193507.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193507.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193507.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
TC0600013537.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193505.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000367361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
ENST00000479265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NM_001001788.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013537.hg.1 |
NR_130110.1 |
Refseq |