Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231347_m1
          See other PHKA2-AS1 GE Assays ›
          Gene Symbol:
          PHKA2-AS1
          Gene Name:
          PHKA2 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 18890296 - 18894975 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231347_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PHKA2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029379.1 2 - 3 468 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG478007.1 - 2 - 3 469 Chr. X - 18890296 - 18894335 [+] on Build GRCh38 74
          GenBankmRNA AW134983.1 - 2 - 3 247 Chr. X - 18892744 - 18894497 [+] on Build GRCh38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein phosphatase with EF-hand domain 1 PPEF1 PP7,
          PPEF,
          PPP7C,
          PPP7CA
          -65.3kb Sense Chr. X - 18675909 - 18827921 on NCBI Build GRCh38
          Downstream uncharacterized LOC101928415 LOC101928415 89.2kb Sense Chr. X - 18984254 - 19082825 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00006729.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000091.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32327811 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11622594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32327809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32327808 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 439500 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 233755 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03665165 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03665163 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21400351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622625 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327847 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622607 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622618 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622659 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327853 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622630 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622645 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622620 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622658 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622629 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622601 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622633 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622651 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622640 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327829 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622636 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622642 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622631 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622641 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327824 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622644 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622648 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622621 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622622 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622599 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622634 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622660 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622607 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622651 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622630 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622621 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622601 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327847 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622634 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622636 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327853 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622660 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622640 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622642 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622618 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327829 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622641 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327824 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622659 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622629 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32327827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622648 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622644 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622622 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622599 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622645 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622631 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622625 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622658 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622633 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622620 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11622596 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03665158 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001235.hg.1 NR_029379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001235.hg.1 TC0X000091.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X001233.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X001233.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X001233.hg.1 NR_029379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001233.hg.1 TC0X000091.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X001234.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X001234.hg.1 NR_029379.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001234.hg.1 TC0X000091.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00146136.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146136.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146136.hg.1 TC0X00006729.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00146136.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146136.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146137.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146137.hg.1 TC0X00006729.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00146137.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 TC0X00006729.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146135.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000439295.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 ENST00000452900.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 NR_029379.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00006729.hg.1 NR_029379.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040731 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040730 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040731 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040730 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 143073 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0