Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231396_m1
          See other LOC389765 GE Assays ›
          Gene Symbol:
          LOC389765
          Gene Name:
          kinesin family member 27 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 85806002 - 85842879 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231396_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC389765
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029410.1 6 - 7 1321 111
          GenBank mRNA AK124523.1 - 3 - 4 785 111
          BC108718.1 - 6 - 7 1253 111
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CK001537.1 - 2 - 3 338 Chr. 9 - 85829507 - 85840844 [+] on Build GRCh38 111

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATP/GTP binding protein 1 AGTPBP1 CCP1,
          NNA1
          -95.7kb Anti-sense Chr. 9 - 85543799 - 85742029 on NCBI Build GRCh38
          Downstream N(alpha)-acetyltransferase 35,
          NatC auxiliary subunit
          NAA35 EGAP,
          MAK10,
          MAK10P,
          bA379P1.1
          100.9kb Sense Chr. 9 - 85941142 - 86026355 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900007768.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000389.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16235100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16235087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16235000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131636 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131652 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131648 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16235080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16235183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16235098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06792043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27131727 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P9280 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17284 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49289 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02124327 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P38779190 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P9280 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P9280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27131583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16235000 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27131557 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06792033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06792034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27131578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27131583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27131578 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16235000 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27131557 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06792033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06792034 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 50556 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 69113 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 44585 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 82900 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16905164 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16901075 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16901694 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16904090 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16903375 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16904954 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02124327 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16899642 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16903505 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16903698 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02124335 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18679408 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16907957 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8162034 - -
          Affymetrix Human Gene 1.0 ST Array 8156165 - -
          Affymetrix Human Gene 1.0 ST Array 8162030 - -
          Affymetrix Human Gene 1.0 ST Array 8162032 - -
          Affymetrix Human Gene 1.0 ST Array 8156166 - -
          Affymetrix Human Gene 1.0 ST Array 8162029 - -
          Affymetrix Human Gene 1.0 ST Array 8162033 - -
          Affymetrix Human Gene 1.0 ST Array 8162036 - -
          Affymetrix Human Gene 1.0 ST Array 8162037 - -
          Affymetrix Human Gene 1.1 ST Array 8156164 - -
          Affymetrix Human Gene 1.1 ST Array 8162019 - -
          Affymetrix HT HG-U133+ PM Array Plate 230815_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR09004171.hg.1 NR_029410.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09004171.hg.1 TC09000389.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09004172.hg.1 NR_029410.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09004172.hg.1 TC09000389.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09004173.hg.1 NR_029410.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09004173.hg.1 TC09000389.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09004170.hg.1 NR_029410.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09004170.hg.1 TC09000389.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900152490.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152490.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152490.hg.1 TC0900007768.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900152490.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152490.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152501.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152501.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152501.hg.1 TC0900007768.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900152501.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152501.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152494.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152494.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152494.hg.1 TC0900007768.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900152494.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152494.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152489.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152489.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152489.hg.1 TC0900007768.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900152489.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152489.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152498.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152498.hg.1 NR_029410.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900152498.hg.1 TC0900007768.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900152498.hg.1 ENST00000431724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900152498.hg.1 NR_029410.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557438_at - -
          Human Genome U133 Plus 2.0 Array 230815_at - -
          Human Genome U133 Plus 2.0 Array 1557438_PM_at - -
          Human Genome U133 Plus 2.0 Array 230815_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230815_at2 - -
          Human Genome U95 Set 77812_at - -
          Human Genome U95 Set 77812_PM_at - -
          Human Genome U95 Set 69263_r_at - -
          Human Genome U95 Set 69263_PM_r_at - -
          Human Genome U133 Set 230815_at - -
          Human Genome U133 Set 230815_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038864 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038864 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3294134 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0