Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231430_sH
          See other MIR30A GE Assays ›
          Gene Symbol:
          MIR30A
          Gene Name:
          microRNA 30a
          Gene Aliases:
          MIRN30A, mir-30a
          Chromosome Location:
          Chr.6: 71403551 - 71403621 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231430_sH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR30A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029504.1 1 41 62
          GenBank mRNA AF480569.1 - 1 45 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 30c-2 MIR30C2 MIRN30C2,
          mir-30c-2
          -26.5kb Sense Chr. 6 - 71376960 - 71377031 on NCBI Build GRCh38
          Downstream long intergenic non-protein coding RNA 472 LINC00472 C6orf155 4.2kb Sense Chr. 6 - 71407864 - 71420745 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0600012261.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001862.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008446 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008488 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008481 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008494 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008484 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008425 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808645 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008457 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008450 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008440 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008470 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008420 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808741 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008480 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008455 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008468 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008461 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808680 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008479 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008453 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008475 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008452 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008464 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008487 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008483 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15829761 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008477 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008459 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808675 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808738 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008489 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008445 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008446 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008461 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008440 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808666 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008480 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008455 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008470 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15829761 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008445 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008450 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008489 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008477 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008479 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008420 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008453 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008487 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008468 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008475 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008484 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008481 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808645 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008459 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008488 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008464 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008457 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808680 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008452 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008494 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008425 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808738 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008491 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008483 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25008493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808741 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04808675 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8127501 - -
          Affymetrix Human Gene 1.1 ST Array 8127500 - -
          Affymetrix Human Transcriptome Array 2.0 TC0600012261.hg.1 ENST00000385092.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012261.hg.1 NR_029504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06022619.hg.1 ENST00000385092.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022619.hg.1 NR_029504.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06022619.hg.1 TC06001862.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600184514.hg.1 ENST00000385092.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600184514.hg.1 NR_029504.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600184514.hg.1 TC0600012261.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600184514.hg.1 NR_029504.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600184514.hg.1 ENST00000385092.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012261.hg.1 ENST00000385092.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012261.hg.1 NR_029504.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308525 NR_029504.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3308525 AF480569.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308525 NR_029504.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308525 AF480569.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308525 NR_029504.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308525 AF480569.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308525 NR_029504.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308525 AF480569.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0