Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231462_s1
          See other MIR181C GE Assays ›
          Gene Symbol:
          MIR181C
          Gene Name:
          microRNA 181c
          Gene Aliases:
          MIRN181C, mir-181c
          Chromosome Location:
          Chr.19: 13874699 - 13874808 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231462_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR181C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029613.1 1 59 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC1900007147.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000244.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Chr array_18 19 21 A_17_P10890854 - -
          Agilent Whole Chr array_18 19 21 A_17_P31506992 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890853 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890876 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890891 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890858 - -
          Agilent Whole Chr array_18 19 21 A_17_P17111444 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890871 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890865 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507040 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890875 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890887 - -
          Agilent Whole Chr array_18 19 21 A_17_P17111446 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890883 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507025 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890868 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890872 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890866 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507042 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890864 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890867 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507004 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890856 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890869 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507001 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890863 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890882 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507009 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507005 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890889 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890870 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890855 - -
          Agilent Whole Chr array_18 19 21 A_17_P17111445 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890873 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890874 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890877 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890881 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890890 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890852 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890892 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890884 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507017 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507018 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507003 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890878 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890888 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507020 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890886 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890862 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507023 - -
          Agilent Whole Chr array_18 19 21 A_17_P31506991 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507016 - -
          Agilent Whole Chr array_18 19 21 A_17_P17111455 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890893 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890885 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890861 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890857 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890860 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890859 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890880 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890893 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890887 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890863 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890861 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890877 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890891 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890885 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507005 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507009 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890892 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890885 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890881 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890877 - -
          Agilent Whole Chr array_18 19 21 A_17_P17111444 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890852 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890879 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890872 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890864 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890876 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890855 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890862 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507016 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890891 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890868 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890884 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890886 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507020 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890861 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890889 - -
          Agilent Whole Chr array_18 19 21 A_17_P17111446 - -
          Agilent Whole Chr array_18 19 21 A_17_P17111445 - -
          Agilent Whole Chr array_18 19 21 A_17_P17111455 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890878 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507017 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507005 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890856 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890874 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507004 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890883 - -
          Agilent Whole Chr array_18 19 21 A_17_P31506992 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890860 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890888 - -
          Agilent Whole Chr array_18 19 21 A_17_P31506991 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507023 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890854 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890870 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890869 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890866 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890873 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890890 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890871 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890887 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507040 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890857 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507042 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890858 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890875 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507025 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890865 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890893 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890880 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890853 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507018 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890882 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890859 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507003 - -
          Agilent Whole Chr array_18 19 21 A_17_P31507001 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890867 - -
          Agilent Whole Chr array_18 19 21 A_17_P10890863 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890877 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890893 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890863 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890891 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890887 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890861 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507004 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890885 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10890858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31507001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8026212 - -
          Affymetrix Human Transcriptome Array 2.0 TC1900007147.hg.1 ENST00000384881.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007147.hg.1 NR_029613.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19003565.hg.1 ENST00000384881.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19003565.hg.1 NR_029613.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19003565.hg.1 TC19000244.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1900007147.hg.1 ENST00000384881.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007147.hg.1 NR_029613.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900151501.hg.1 ENST00000384881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900151501.hg.1 NR_029613.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900151501.hg.1 TC1900007147.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900151501.hg.1 NR_029613.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900151501.hg.1 ENST00000384881.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3310201 NR_029613.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3310201 NR_029613.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3310201 NR_029613.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3310201 NR_029613.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0