Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231473_s1
          See other MIR181A1 GE Assays ›
          Gene Symbol:
          MIR181A1
          Gene Name:
          microRNA 181a-1
          Gene Aliases:
          MIR213, MIRN181A1, MIRN213, hsa-mir-181a-1, mir-181a-1, mir-213
          Chromosome Location:
          Chr.1: 198859044 - 198859153 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231473_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR181A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029626.1 1 57 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream MIR181A1 host gene MIR181A1HG Sense Chr. 1 - 198808003 - 198937429 on NCBI Build GRCh38
          Downstream long intergenic non-protein coding RNA 1222 LINC01222 TCONS_00000112 133.4kb Sense Chr. 1 - 198992589 - 199018964 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0100016865.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003672.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776109 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716492 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776132 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776119 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716490 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776120 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716484 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776106 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716499 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776140 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716484 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776120 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776119 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776109 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776106 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716499 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716492 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776132 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15159290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716490 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00716474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20776104 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 7923175 - -
          Affymetrix Human Transcriptome Array 2.0 TC0100016865.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016865.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016865.hg.1 NR_029626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016865.hg.1 NR_029626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01056285.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01056285.hg.1 NR_029626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01056285.hg.1 TC01003672.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100227600.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100227600.hg.1 NR_029626.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100227600.hg.1 TC0100016865.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100227600.hg.1 NR_029626.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100227600.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100227601.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100227601.hg.1 NR_029626.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100227601.hg.1 TC0100016865.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100227601.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100227601.hg.1 NR_029626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016865.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016865.hg.1 ENST00000385026.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016865.hg.1 NR_029626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016865.hg.1 NR_029626.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009362 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009362 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309289 NR_029626.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309289 NR_029626.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309289 NR_029626.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309289 NR_029626.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0