Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231530_s1
          See other MIR194-1 GE Assays ›
          Gene Symbol:
          MIR194-1
          Gene Name:
          microRNA 194-1
          Gene Aliases:
          MIRN194-1, mir-194-1
          Chromosome Location:
          Chr.1: 220118157 - 220118241 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231530_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR194-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029711.1 1 42 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 215 MIR215 MIRN215,
          miRNA215,
          mir-215
          -195.0 Sense Chr. 1 - 220117853 - 220117962 on NCBI Build GRCh38
          Downstream RAB3 GTPase activating non-catalytic protein subunit 2 RAB3GAP2 RAB3-GAP150,
          RAB3GAP150,
          SPG69,
          WARBM2,
          p150
          30.0kb Sense Chr. 1 - 220148268 - 220272501 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0100017372.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003837.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879735 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879738 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879729 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879734 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15176320 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879724 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817271 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879723 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879716 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879738 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879734 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15176320 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879713 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879729 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879724 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879735 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879746 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20879722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00817271 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 7924403 - -
          Affymetrix Human Transcriptome Array 2.0 PSR01058628.hg.1 ENST00000384892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01058628.hg.1 NR_029711.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01058628.hg.1 TC01003837.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100017372.hg.1 ENST00000384892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017372.hg.1 NR_029711.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017372.hg.1 ENST00000384892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017372.hg.1 NR_029711.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100231459.hg.1 ENST00000384892.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100231459.hg.1 NR_029711.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100231459.hg.1 TC0100017372.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100231459.hg.1 NR_029711.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100231459.hg.1 ENST00000384892.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309478 NR_029711.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309478 NR_029711.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309478 NR_029711.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309478 NR_029711.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0