Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04231662_s1
          See other MIR532 GE Assays ›
          Gene Symbol:
          MIR532
          Gene Name:
          microRNA 532
          Gene Aliases:
          MIRN532, hsa-mir-532, mir-532
          Chromosome Location:
          Chr.X: 50003148 - 50003238 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04231662_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR532
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030241.2 1 42 59

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ubiquitin specific peptidase 27,
          X-linked
          USP27X USP22L,
          USP27
          -120.6kb Sense Chr. X - 49879867 - 49882565 on NCBI Build GRCh38
          Downstream microRNA 188 MIR188 MIRN188,
          miR-188
          265.0 Sense Chr. X - 50003503 - 50003588 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0X00007241.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000279.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453549 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453568 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453528 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745163 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453553 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453551 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745170 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453572 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453563 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453530 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453554 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453532 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453545 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453552 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453555 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453547 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745132 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453526 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453538 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453547 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745145 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453552 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453532 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453553 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453576 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745163 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745170 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453554 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745132 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453545 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453551 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453528 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453530 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453544 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453568 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453572 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453555 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11745124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453563 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453538 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453549 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32453526 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00007241.hg.1 ENST00000385025.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00007241.hg.1 NR_030241.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X003846.hg.1 ENST00000385025.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X003846.hg.1 NR_030241.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X003846.hg.1 TC0X000279.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0X00007241.hg.1 ENST00000385025.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00007241.hg.1 NR_030241.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00149907.hg.1 ENST00000385025.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00149907.hg.1 NR_030241.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00149907.hg.1 TC0X00007241.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00149907.hg.1 NR_030241.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00149907.hg.1 ENST00000385025.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308647 NR_030241.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308647 NR_030241.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308647 NR_030241.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308647 NR_030241.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0