Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009026.hg.1 |
ENST00000585264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009026.hg.1 |
NR_037507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010823.hg.1 |
NR_037507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010823.hg.1 |
TC15001206.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500009026.hg.1 |
ENST00000585264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009026.hg.1 |
NR_037507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500165224.hg.1 |
ENST00000585264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500165224.hg.1 |
NR_037507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500165224.hg.1 |
TC1500009026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500165224.hg.1 |
NR_037507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500165224.hg.1 |
ENST00000585264.1 |
Ensembl |