Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04232235_m1
          See other HHIP-AS1 GE Assays ›
          Gene Symbol:
          HHIP-AS1
          Gene Name:
          HHIP antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 144642916 - 144646019 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04232235_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HHIP-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037595.1 1 - 2 316 95
          GenBank mRNA BC039450.1 - 1 - 2 319 95
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI667372.1 - 1 - 2 324 Chr. 4 - 144643321 - 144646026 [-] on Build GRCh38 95
          GenBankmRNA DB452221.1 - 1 - 2 230 Chr. 4 - 144643464 - 144645933 [-] on Build GRCh38 95
          GenBankmRNA BI465314.1 - 2 - 3 233 Chr. 4 - 144643305 - 144645984 [-] on Build GRCh38 94
          GenBankmRNA BI465333.1 - 236 Chr. - - [-] on Build 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream hedgehog interacting protein HHIP HIP Anti-sense Chr. 4 - 144645996 - 144742645 on NCBI Build GRCh38
          Downstream anaphase promoting complex subunit 10 ANAPC10 APC10,
          DOC1
          321.1kb Sense Chr. 4 - 144967117 - 145098541 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400012021.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001607.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03496661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03496659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23658644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03496667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03496660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23658639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23658633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23658642 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 140068 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 492151 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P115050 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P115050 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P36947546 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 172221 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P36947546 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01097538 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36947546 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 236632_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR04022435.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04022435.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04022435.hg.1 TC04001607.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04022436.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04022436.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04022436.hg.1 TC04001607.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04022440.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04022440.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04022440.hg.1 TC04001607.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04022437.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04022437.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04022437.hg.1 TC04001607.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04022439.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04022439.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04022439.hg.1 TC04001607.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04022441.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04022441.hg.1 TC04001607.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182508.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182508.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182508.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182508.hg.1 TC0400012021.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400182508.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182508.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182508.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182510.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182510.hg.1 TC0400012021.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400182510.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182505.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182505.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182505.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182505.hg.1 TC0400012021.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400182505.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182505.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182505.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 TC0400012021.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182506.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182509.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182509.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182509.hg.1 TC0400012021.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400182509.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182509.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182504.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182504.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182504.hg.1 TC0400012021.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400182504.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182504.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000503066.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000508269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 ENST00000512359.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 NR_037595.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012021.hg.1 NR_037595.1 Refseq
          Human Genome U133 Plus 2.0 Array 236632_PM_at - -
          Human Genome U133 Plus 2.0 Array 236632_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236632_at5 - -
          Human Genome U95 Set 46938_at - -
          Human Genome U95 Set 46938_PM_at - -
          Human Genome U133 Set 236632_at - -
          Human Genome U133 Set 236632_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024808 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024808 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 143230 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3288032 NR_037595.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3288032 BC039450.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3288032 CR620567.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0