Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04232304_m1
          See other LOC100287010 GE Assays ›
          Gene Symbol:
          LOC100287010
          Gene Name:
          uncharacterized LOC100287010
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 104378850 - 104408332 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04232304_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100287010
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037885.2 2 - 3 191 79
          GenBank mRNA AK296254.1 - 2 - 3 199 79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DC341463.1 - 2 - 3 199 Chr. 2 - 104379354 - 104408332 [-] on Build GRCh38 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105373519 LOC105373519 -1.4mb Sense Chr. 2 - 102967408 - 102984429 on NCBI Build GRCh38
          Downstream uncharacterized LOC101927331 LOC101927331 18.2kb Sense Chr. 2 - 104412152 - 104414031 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013723.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002156.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK296254.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 84970 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 506991 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 921925 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702964 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 772665 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 747604 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 918665 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 885371 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 927039 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 771780 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 280308 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P35794474 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P35794474 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P13334767 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35794474 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15766555 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02035163.hg.1 NR_037885.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035163.hg.1 TC02002156.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02035164.hg.1 NR_037885.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035164.hg.1 TC02002156.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02035162.hg.1 NR_037885.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035162.hg.1 TC02002156.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200200596.hg.1 NR_037885.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200200596.hg.1 TC0200013723.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200200596.hg.1 NR_037885.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200200598.hg.1 NR_037885.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200200598.hg.1 TC0200013723.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200200598.hg.1 NR_037885.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200200597.hg.1 NR_037885.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200200597.hg.1 TC0200013723.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200200597.hg.1 NR_037885.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b776c6558-ktt9x:80/100.66.134.75:80.
          git-commit: 08b7be5c3d3493be030128aff2ceec7cf49358c5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc